Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
ENST00000441882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
ENST00000441882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
ENST00000441882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
ENST00000448513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
ENST00000448513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
ENST00000448513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
NR_120519.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
NR_120519.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
NR_120519.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
NR_120520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
NR_120520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012031.hg.1 |
NR_120520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186472.hg.1 |
ENST00000441882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186472.hg.1 |
ENST00000448513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186472.hg.1 |
NR_120519.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186472.hg.1 |
NR_120520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186472.hg.1 |
TC0700012031.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700186472.hg.1 |
ENST00000441882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186472.hg.1 |
ENST00000448513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186472.hg.1 |
NR_120519.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186472.hg.1 |
NR_120520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186473.hg.1 |
ENST00000441882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186473.hg.1 |
ENST00000448513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186473.hg.1 |
NR_120519.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186473.hg.1 |
NR_120520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186473.hg.1 |
TC0700012031.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700186473.hg.1 |
ENST00000441882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186473.hg.1 |
ENST00000448513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186473.hg.1 |
NR_120519.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186473.hg.1 |
NR_120520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
ENST00000441882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
ENST00000441882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
ENST00000441882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
ENST00000448513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
ENST00000448513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
ENST00000448513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
NR_120519.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
NR_120519.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
NR_120519.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
NR_120520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
NR_120520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012031.hg.1 |
NR_120520.1 |
Refseq |