Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01550814_m1
          See other MX2 GE Assays ›
          Gene Symbol:
          MX2
          Gene Name:
          MX dynamin like GTPase 2
          Gene Aliases:
          MXB
          Chromosome Location:
          Chr.21: 41361986 - 41408943 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01550814_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MX2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002463.1 NP_002454.1 8 - 9 1259 72
          XM_005260983.4 XP_005261040.1 8 - 9 1357 72
          XM_005260984.1 XP_005261041.1 8 - 9 1537 72
          XM_011529571.1 XP_011527873.1 8 - 9 1292 72
          XM_011529572.1 XP_011527874.1 8 - 9 1395 72
          XM_011529573.1 XP_011527875.1 7 - 8 1161 72
          XM_011529574.2 XP_011527876.1 3 - 4 439 72
          GenBank mRNA AK122952.1 - 3 - 4 1445 72
          AK298780.1 - 6 - 7 1217 72
          BC035293.1 - 7 - 8 1221 5182160 72
          M30818.1 - 8 - 9 1259 72
          M33883.1 - 7 - 8 1072 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100007205.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000188.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MX2

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          response to virus
          protein transport
          response to interferon-alpha
          regulation of nucleocytoplasmic transport
          mRNA transport
          defense response to virus
          regulation of cell cycle
          type I interferon signaling pathway
          GTPase activity
          protein binding
          GTP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000330714.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000398632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000398632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000398632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000398632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000398632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000398632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000398632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000398632.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000416447.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000416447.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000416447.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000416447.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000416447.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000416447.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000418103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000418103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000418103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000418103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000418103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000435611.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000435611.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000435611.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000435611.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000435611.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000435611.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000436410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000436410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000436410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000436410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000436410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000436410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000474368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000474368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000474368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000474368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000474368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000481838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000481838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000481838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000481838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000481838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000482953.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000493753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000493753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000493753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000493753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000493753.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000494252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000494252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000494252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000494252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000495892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000495892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000495892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000495892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000495892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000495892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000496774.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000496774.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000496774.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000496774.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000496774.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000496774.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000496774.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 ENST00000496774.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007205.hg.1 NM_002463.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0