Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01551502_m1
          See other MBL2 GE Assays ›
          Gene Symbol:
          MBL2
          Gene Name:
          mannose binding lectin 2
          Gene Aliases:
          COLEC1, HSMBPC, MBL, MBL2D, MBP, MBP-C, MBP1, MBPD
          Chromosome Location:
          Chr.10: 52764977 - 52772845 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01551502_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MBL2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000242.2 NP_000233.1 2 - 3 370 69
          XM_006717861.3 XP_006717924.1 3 - 4 438 69
          XM_011539816.2 XP_011538118.1 3 - 4 422 69
          GenBank mRNA AF360991.1 - 2 - 3 321 69
          AY826184.1 - 2 - 3 305 69
          BC069338.1 - 2 - 3 355 7262231 69
          BC096179.3 - 2 - 3 336 40003735 69
          BC096180.3 - 2 - 3 336 40003741 69
          BC096181.3 - 2 - 3 336 40003742 69
          BC096182.3 - 2 - 3 336 40003743 69
          X15422.1 - 2 - 3 370 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000010661.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001301.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MBL2

          Gene Ontology Categories:

          Function(s) Process(es)

          complement activation, lectin pathway
          proteolysis
          acute-phase response
          complement activation
          complement activation, classical pathway
          response to oxidative stress
          opsonization
          defense response to bacterium
          innate immune response
          negative regulation of viral process
          positive regulation of phagocytosis
          defense response to Gram-positive bacterium
          killing by host of symbiont cells
          serine-type endopeptidase activity
          signaling receptor binding
          calcium ion binding
          protein binding
          mannose binding
          calcium-dependent protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10016267.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016267.hg.1 ENST00000493043.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016267.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10016267.hg.1 TC10001301.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000172707.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000172707.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000172707.hg.1 TC1000010661.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000172707.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000172707.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 ENST00000373968.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010661.hg.1 NM_000242.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0