Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008012.hg.1 |
ENST00000339507.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008012.hg.1 |
ENST00000348497.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008012.hg.1 |
ENST00000441236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008012.hg.1 |
ENST00000449359.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008012.hg.1 |
NM_001184824.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008012.hg.1 |
NM_019023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008012.hg.1 |
TC16000555.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000339507.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000441236.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000449359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000562050.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000563520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000564050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000564441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000565356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000567542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000568975.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000569571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
NM_001184824.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
NM_001290018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
NM_019023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
TC1600008193.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000562050.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000339507.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000441236.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000449359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000563520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000564050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000564441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000565356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000567542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000568975.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
ENST00000569571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
NM_001184824.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
NM_001290018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161388.hg.1 |
NM_019023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000339507.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000441236.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000449359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000562050.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000562381.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000563520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000564050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000564441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000565356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000568975.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000569047.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000569571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
NM_001184824.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
NM_001290018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
NM_019023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
TC1600008193.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000339507.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000441236.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000449359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000562050.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000562381.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000563520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000564050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000564441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000565356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000568975.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000569047.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
ENST00000569571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
NM_001184824.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
NM_001290018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161386.hg.1 |
NM_019023.3 |
Refseq |