Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01553013_g1
          See other VEZT GE Assays ›
          Gene Symbol:
          VEZT
          Gene Name:
          vezatin, adherens junctions transmembrane protein
          Gene Aliases:
          VEZATIN
          Chromosome Location:
          Chr.12: 95217746 - 95302799 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01553013_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: VEZT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017599.3 NP_060069.3 7 - 8 1097 86
          NR_038241.1 8 - 9 1170 86
          NR_038242.1 7 - 8 1105 86
          XM_006719477.3 XP_006719540.1 7 - 8 1109 86
          XM_006719478.2 XP_006719541.1 6 - 7 1003 86
          XM_006719479.2 XP_006719542.1 8 - 9 1180 86
          XM_006719480.3 XP_006719543.1 8 - 9 1157 86
          XM_006719481.2 XP_006719544.1 7 - 8 1027 86
          XM_006719482.3 XP_006719545.1 7 - 8 1408 86
          XM_006719483.3 XP_006719546.1 7 - 8 1425 86
          XM_006719484.3 XP_006719547.1 6 - 7 1335 86
          XM_006719485.2 XP_006719548.1 5 - 6 777 86
          XM_006719486.3 XP_006719549.1 5 - 6 765 86
          XM_006719490.3 XP_006719553.1 8 - 9 1168 86
          XM_011538543.1 XP_011536845.1 8 - 9 1102 86
          XM_011538544.1 XP_011536846.1 8 - 9 1090 86
          XM_011538545.2 XP_011536847.1 7 - 8 1070 86
          XM_011538546.1 XP_011536848.1 9 - 10 1173 86
          XM_011538547.1 XP_011536849.1 9 - 10 1217 86
          XM_011538548.2 XP_011536850.1 6 - 7 863 86
          XM_011538549.1 XP_011536851.1 8 - 9 1102 86
          XM_011538550.2 XP_011536852.1 8 - 9 1102 86
          XM_011538551.2 XP_011536853.1 8 - 9 1090 86
          XM_011538552.1 XP_011536854.1 8 - 9 1176 86
          XM_011538553.2 XP_011536855.1 8 - 9 1106 86
          XM_017019586.1 XP_016875075.1 7 - 8 1058 86
          XM_017019587.1 XP_016875076.1 6 - 7 991 86
          XM_017019588.1 XP_016875077.1 9 - 10 1161 86
          XM_017019589.1 XP_016875078.1 8 - 9 1145 86
          XM_017019590.1 XP_016875079.1 9 - 10 1205 86
          XM_017019591.1 XP_016875080.1 7 - 8 1396 86
          XM_017019592.1 XP_016875081.1 6 - 7 850 86
          XM_017019593.1 XP_016875082.1 9 - 10 1225 86
          XM_017019594.1 XP_016875083.1 8 - 9 1087 86
          XM_017019595.1 XP_016875084.1 7 - 8 1109 86
          XM_017019596.1 XP_016875085.1 7 - 8 1109 86
          XM_017019597.1 XP_016875086.1 7 - 8 1097 86
          XM_017019598.1 XP_016875087.1 7 - 8 1097 86
          XM_017019599.1 XP_016875088.1 7 - 8 1058 86
          XM_017019600.1 XP_016875089.1 7 - 8 1413 86
          XM_017019601.1 XP_016875090.1 8 - 9 1164 86
          XM_017019602.1 XP_016875091.1 9 - 10 1161 86
          XM_017019603.1 XP_016875092.1 7 - 8 1425 86
          XM_017019604.1 XP_016875093.1 5 - 6 777 86
          XM_017019605.1 XP_016875094.1 5 - 6 765 86
          GenBank mRNA AF216644.1 - 7 - 8 1024 86
          AF277625.1 - 3 - 4 536 86
          AK001338.1 - 5 - 6 778 86
          AK225303.1 - 8 - 9 1110 86
          AK225356.1 - 8 - 9 1119 86
          AK307649.1 - 8 - 9 1176 86
          AK307864.1 - 5 - 6 764 86
          AK309943.1 - 7 - 8 1044 86
          AL137497.1 - 8 - 9 1092 86
          BC028597.1 - 10 - 11 1426 86
          BC064939.1 - 8 - 9 1087 6159768 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene VEZT

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-cell adhesion
          myosin binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix