Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01553881_g1
          See other SIPA1L1 GE Assays ›
          Gene Symbol:
          SIPA1L1
          Gene Name:
          signal induced proliferation associated 1 like 1
          Gene Aliases:
          E6TP1
          Chromosome Location:
          Chr.14: 71320418 - 71741229 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01553881_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SIPA1L1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001284247.1 NP_001271176.1 12 - 13 4010 68
          NM_015556.2 NP_056371.1 13 - 14 4069 68
          XM_005267516.4 XP_005267573.1 13 - 14 4658 68
          XM_006720111.2 XP_006720174.1 15 - 16 5948 68
          XM_011536630.1 XP_011534932.1 17 - 18 4420 68
          XM_011536631.1 XP_011534933.1 17 - 18 4831 68
          XM_011536632.1 XP_011534934.1 16 - 17 4340 68
          XM_011536635.2 XP_011534937.1 16 - 17 4487 68
          XM_011536638.2 XP_011534940.1 12 - 13 3143 68
          XM_017021159.1 XP_016876648.1 19 - 20 4577 68
          XM_017021160.1 XP_016876649.1 17 - 18 4499 68
          XM_017021161.1 XP_016876650.1 18 - 19 4452 68
          XM_017021162.1 XP_016876651.1 20 - 21 4634 68
          XM_017021163.1 XP_016876652.1 15 - 16 4301 68
          XM_017021164.1 XP_016876653.1 15 - 16 4308 68
          XM_017021165.1 XP_016876654.1 17 - 18 4377 68
          XM_017021166.1 XP_016876655.1 18 - 19 4510 68
          XM_017021167.1 XP_016876656.1 17 - 18 4385 68
          XM_017021168.1 XP_016876657.1 17 - 18 4359 68
          XM_017021169.1 XP_016876658.1 16 - 17 4512 68
          XM_017021170.1 XP_016876659.1 16 - 17 4870 68
          XM_017021171.1 XP_016876660.1 17 - 18 4443 68
          XM_017021172.1 XP_016876661.1 16 - 17 4318 68
          XM_017021173.1 XP_016876662.1 15 - 16 4445 68
          XM_017021174.1 XP_016876663.1 15 - 16 4685 68
          XM_017021175.1 XP_016876664.1 15 - 16 4803 68
          XM_017021176.1 XP_016876665.1 20 - 21 4630 68
          XM_017021177.1 XP_016876666.1 20 - 21 4619 68
          XM_017021178.1 XP_016876667.1 18 - 19 4457 68
          XM_017021179.1 XP_016876668.1 17 - 18 4360 68
          XM_017021180.1 XP_016876669.1 21 - 22 4672 68
          XM_017021181.1 XP_016876670.1 19 - 20 4577 68
          XM_017021182.1 XP_016876671.1 18 - 19 4510 68
          XM_017021183.1 XP_016876672.1 16 - 17 4318 68
          XM_017021184.1 XP_016876673.1 20 - 21 4630 68
          XM_017021185.1 XP_016876674.1 13 - 14 4658 68
          XM_017021194.1 XP_016876683.1 18 - 19 4430 68
          XM_017021197.1 XP_016876686.1 18 - 19 2777 68
          XM_017021198.1 XP_016876687.1 15 - 16 2518 68
          XM_017021199.1 XP_016876688.1 19 - 20 2819 68
          GenBank mRNA AB007900.2 - 12 - 13 3793 68
          AF090990.1 - 13 - 14 4056 68
          BC146777.1 - 12 - 13 3793 9007226 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400007595.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000433.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SIPA1L1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          actin cytoskeleton reorganization
          regulation of GTPase activity
          ephrin receptor signaling pathway
          regulation of synaptic plasticity
          regulation of axonogenesis
          regulation of small GTPase mediated signal transduction
          regulation of dendritic spine morphogenesis
          activation of GTPase activity
          molecular_function
          GTPase activator activity
          ephrin receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14005213.hg.1 ENST00000358550.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14005213.hg.1 ENST00000381232.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14005213.hg.1 ENST00000537413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14005213.hg.1 ENST00000554874.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14005213.hg.1 ENST00000555818.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14005213.hg.1 NM_015556.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14005213.hg.1 TC14000433.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000358550.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000381232.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000537413.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000554874.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000554960.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000555818.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 NM_001284245.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 NM_001284246.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 NM_001284247.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 NM_015556.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 TC1400007595.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000358550.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000381232.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000537413.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000554874.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000554960.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 ENST00000555818.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 NM_001284245.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 NM_001284246.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 NM_001284247.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400153372.hg.1 NM_015556.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0