Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017544.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017544.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017544.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017544.hg.1 |
TC09001406.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900174813.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174813.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174813.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174813.hg.1 |
TC0900010975.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900174813.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174813.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174813.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000259455.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
ENST00000477471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010975.hg.1 |
NM_005458.7 |
Refseq |