Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01556136_m1
          See other BMPR2 GE Assays ›
          Gene Symbol:
          BMPR2
          Gene Name:
          bone morphogenetic protein receptor type 2
          Gene Aliases:
          BMPR-II, BMPR3, BMR2, BRK-3, POVD1, PPH1, T-ALK
          Chromosome Location:
          Chr.2: 202376310 - 202567751 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01556136_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BMPR2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001204.6 NP_001195.2 8 - 9 2270 113
          XM_011511687.1 XP_011509989.1 8 - 9 2287 113
          GenBank mRNA AK292430.1 - 8 - 9 1534 113
          BC052985.2 - 8 - 9 1696 6149698 113
          D50516.1 - 8 - 9 1530 113
          U20165.1 - 8 - 9 1158 113
          Z48923.1 - 8 - 9 1503 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200010489.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001192.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BMPR2

          Gene Ontology Categories:

          Function(s) Process(es)

          mesoderm formation
          maternal placenta development
          endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lymphangiogenesis
          blood vessel remodeling
          chondrocyte development
          negative regulation of systemic arterial blood pressure
          transcription by RNA polymerase II
          transmembrane receptor protein serine/threonine kinase signaling pathway
          brain development
          cellular response to starvation
          anterior/posterior pattern specification
          positive regulation of endothelial cell migration
          positive regulation of epithelial cell migration
          positive regulation of pathway-restricted SMAD protein phosphorylation
          regulation of lung blood pressure
          proteoglycan biosynthetic process
          negative regulation of cell growth
          positive regulation of bone mineralization
          BMP signaling pathway
          positive regulation of BMP signaling pathway
          activin receptor signaling pathway
          regulation of cell population proliferation
          positive regulation of osteoblast differentiation
          negative regulation of vasoconstriction
          positive regulation of transcription by RNA polymerase II
          vascular endothelial growth factor receptor signaling pathway
          lung alveolus development
          positive regulation of axon extension involved in axon guidance
          limb development
          endochondral bone morphogenesis
          lymphatic endothelial cell differentiation
          artery development
          venous blood vessel development
          positive regulation of cartilage development
          retina vasculature development in camera-type eye
          cellular response to BMP stimulus
          endothelial cell apoptotic process
          negative regulation of chondrocyte proliferation
          negative regulation of DNA biosynthetic process
          protein binding
          ATP binding
          activin receptor activity, type II
          growth factor binding
          BMP binding
          metal ion binding
          BMP receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000479069.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000479069.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 ENST00000479069.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018142.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018142.hg.1 ENST00000374580.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018142.hg.1 NM_001204.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018142.hg.1 TC02001192.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200174211.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174211.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174211.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay PSR0200174211.hg.1 TC0200010489.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200174211.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay PSR0200174211.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174211.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374574.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000374580.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000479069.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000479069.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 ENST00000479069.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq
          Affymetrix Human Clariom D Assay TC0200010489.hg.1 NM_001204.6 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix