Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000557836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000557836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
ENST00000557836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026150.hg.1 |
ENST00000261731.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026150.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026150.hg.1 |
TC12001990.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000557836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000557836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
ENST00000557836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011996.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193456.hg.1 |
ENST00000261731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193456.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193456.hg.1 |
TC1200011996.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200193456.hg.1 |
NM_022363.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193456.hg.1 |
ENST00000261731.3 |
Ensembl |