Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01585981_s1
          See other LOC153811 GE Assays ›
          Gene Symbol:
          LOC153811 RNF130
          Gene Name:
          uncharacterized LOC153811
          ring finger protein 130
          Gene Aliases:
          -
          G1RZFP, GOLIATH, GP
          Chromosome Location:
          -
          Chromosome Location:
          Chr.5: 179911651 - 180072130 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01585981_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC153811
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK021734.1 - 1 764 112
          Gene Symbol: RNF130
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011534593.2 XP_011532895.1 8 6259 112
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA786152.1 - 1 459 Chr. 5 - 179915686 - 179916203 [-] on Build GRCh38 112
          GenBankmRNA DA783748.1 - 1 442 Chr. 5 - 179915603 - 179916187 [-] on Build GRCh38 112
          GenBankmRNA DA496392.1 - 1 452 Chr. 5 - 179915619 - 179916196 [-] on Build GRCh38 112

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream TBC1 domain family member 9B TBC1D9B GRAMD9B -7.7kb Sense Chr. 5 - 179862071 - 179907856 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          programmed cell death
          protein ubiquitination
          ubiquitin-protein transferase activity
          zinc ion binding
          ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680112 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04495894 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04495899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04495892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04495897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04495891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04495889 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680109 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680102 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680082 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04495902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04495876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04495901 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24680088 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 1682 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 168797 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 238698 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 372559 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3671506 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P614261 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 26932 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 24077 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01405282 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01405282 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3671506 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P614261 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P01405282 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01405282 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P614261 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 85539 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 923 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01405282 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01405280 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2155105 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2155105 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01405282 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 232192_PM_at - -
          Human Genome U133 Plus 2.0 Array 232192_PM_at - -
          Human Genome U133 Plus 2.0 Array 232192_at - -
          Human Genome U133 Plus 2.0 Array 1559537_at - -
          Human Genome U133 Plus 2.0 Array 1559537_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232192_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559537_at7 - -
          Human Genome U95 Set 59208_at - -
          Human Genome U95 Set 59208_PM_at - -
          Human Genome U133 Set 232192_PM_at - -
          Human Genome U133 Set 232192_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 228735 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1897836 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 2690731 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_114805 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1833040 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1833040 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1897836 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0