Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01587197_m1
          See other ARNTL GE Assays ›
          Gene Symbol:
          ARNTL
          Gene Name:
          aryl hydrocarbon receptor nuclear translocator like
          Gene Aliases:
          BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
          Chromosome Location:
          Chr.11: 13276613 - 13387268 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01587197_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ARNTL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030272.2 NP_001025443.1 6 - 7 628 136
          NM_001030273.2 NP_001025444.1 7 - 8 699 136
          NM_001178.5 NP_001169.3 7 - 8 683 136
          NM_001297719.1 NP_001284648.1 7 - 8 683 136
          NM_001297722.1 NP_001284651.1 6 - 7 628 136
          NM_001297724.1 NP_001284653.1 7 - 8 692 136
          XM_006718233.3 XP_006718296.1 6 - 7 570 136
          XM_006718234.1 XP_006718297.1 7 - 8 636 136
          XM_011520105.2 XP_011518407.2 5 - 6 734 136
          XM_011520107.2 XP_011518409.2 5 - 6 729 136
          XM_011520108.1 XP_011518410.1 7 - 8 676 136
          XM_011520109.1 XP_011518411.1 5 - 6 431 136
          XM_011520110.1 XP_011518412.1 8 - 9 691 136
          XM_011520111.1 XP_011518413.1 6 - 7 506 136
          XM_011520112.2 XP_011518414.1 8 - 9 691 136
          XM_011520113.1 XP_011518415.1 8 - 9 691 136
          XM_017017738.1 XP_016873227.1 5 - 6 732 136
          XM_017017739.1 XP_016873228.1 5 - 6 728 136
          XM_017017740.1 XP_016873229.1 6 - 7 570 136
          XM_017017741.1 XP_016873230.1 5 - 6 431 136
          XM_017017742.1 XP_016873231.1 5 - 6 431 136
          XM_017017743.1 XP_016873232.1 5 - 6 431 136
          XM_017017744.1 XP_016873233.1 8 - 9 691 136
          XM_017017745.1 XP_016873234.1 7 - 8 636 136
          XM_017017746.1 XP_016873235.1 7 - 8 646 136
          XM_017017747.1 XP_016873236.1 6 - 7 506 136
          XM_017017748.1 XP_016873237.1 6 - 7 563 136
          GenBank mRNA AB000812.1 - 4 - 5 264 136
          AB000815.1 - 5 - 6 335 136
          AB000816.1 - 6 - 7 675 136
          AF044288.1 - 7 - 8 593 136
          AK095749.1 - 7 - 8 584 136
          AK291510.1 - 6 - 7 541 136
          AK315174.1 - 7 - 8 621 136
          BC016674.1 - 4 - 5 227 3921870 136
          BC031214.1 - 7 - 8 524 4793007 136
          BC041129.1 - 7 - 8 607 5753529 136
          D89722.1 - 4 - 5 240 136
          U60415.1 - 6 - 7 527 136

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ARNTL

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          spermatogenesis
          circadian rhythm
          negative regulation of TOR signaling
          circadian regulation of gene expression
          regulation of protein catabolic process
          regulation of hair cycle
          positive regulation of circadian rhythm
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of fat cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          regulation of neurogenesis
          regulation of insulin secretion
          regulation of cell cycle
          response to redox state
          maternal process involved in parturition
          positive regulation of canonical Wnt signaling pathway
          oxidative stress-induced premature senescence
          regulation of type B pancreatic cell development
          negative regulation of glucocorticoid receptor signaling pathway
          regulation of cellular senescence
          positive regulation of skeletal muscle cell differentiation
          transcription cis-regulatory region binding
          core promoter sequence-specific DNA binding
          DNA binding
          protein binding
          aryl hydrocarbon receptor binding
          bHLH transcription factor binding
          sequence-specific DNA binding
          protein heterodimerization activity
          Hsp90 protein binding
          E-box binding
          DNA-binding transcription factor binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0