Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013469.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013469.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013469.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013469.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013469.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013469.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013469.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013469.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013469.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013469.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013469.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013469.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013469.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013469.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013469.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013469.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013469.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013469.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013469.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013469.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193068.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193068.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193068.hg.1 |
TC0600013469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193068.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193068.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193069.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193069.hg.1 |
NR_125860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193069.hg.1 |
TC0600013469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193069.hg.1 |
ENST00000419168.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193069.hg.1 |
NR_125860.1 |
Refseq |