Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01597796_m1
          See other CRY1 GE Assays ›
          Gene Symbol:
          CRY1
          Gene Name:
          cryptochrome circadian clock 1
          Gene Aliases:
          PHLL1
          Chromosome Location:
          Chr.12: 106991364 - 107093872 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01597796_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CRY1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004075.4 NP_004066.1 10 - 11 2477 72
          XM_017018832.1 XP_016874321.1 11 - 12 2612 72
          GenBank mRNA AK098615.1 - 5 - 6 881 72
          AK125915.1 - 12 - 13 2160 72
          AK290552.1 - 10 - 11 1777 72
          BC017466.1 - 10 - 11 1900 72
          BC030519.1 - 10 - 11 2150 5169081 72
          D83702.1 - 10 - 11 2167 72
          D84657.1 - 10 - 11 1581 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200011812.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001925.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CRY1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          gluconeogenesis
          transcription, DNA-templated
          DNA damage induced protein phosphorylation
          blue light signaling pathway
          protein-chromophore linkage
          lipid storage
          negative regulation of protein ubiquitination
          response to insulin
          circadian regulation of gene expression
          response to glucagon
          glucose homeostasis
          regulation of circadian rhythm
          negative regulation of circadian rhythm
          entrainment of circadian clock by photoperiod
          negative regulation of G protein-coupled receptor signaling pathway
          negative regulation of transcription, DNA-templated
          regulation of DNA damage checkpoint
          negative regulation of glucocorticoid receptor signaling pathway
          negative regulation of glucocorticoid secretion
          nucleotide binding
          core promoter sequence-specific DNA binding
          DNA binding
          double-stranded DNA binding
          deoxyribodipyrimidine photo-lyase activity
          DNA (6-4) photolyase activity
          protein binding
          transcription factor binding
          blue light photoreceptor activity
          protein kinase binding
          phosphatase binding
          histone deacetylase binding
          ubiquitin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12025084.hg.1 ENST00000008527.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12025084.hg.1 ENST00000319645.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12025084.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12025084.hg.1 TC12001925.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12025085.hg.1 ENST00000008527.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12025085.hg.1 ENST00000319645.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12025085.hg.1 NM_004075.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12025085.hg.1 TC12001925.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200191524.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200191524.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200191524.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200191524.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200191524.hg.1 TC1200011812.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200191524.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200191524.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200191524.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200191524.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000008527.9 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000546722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000549356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000550633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 ENST00000552790.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011812.hg.1 NM_004075.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0