Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01687541_s1
          See other LOC392196 GE Assays ›
          Gene Symbol:
          LOC392196
          Gene Name:
          ubiquitin specific peptidase 17-like family member 2 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 12127858 - 12129297 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01687541_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC392196
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003275.2 1 332 155

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream family with sequence similarity 66 member D FAM66D Anti-sense Chr. 8 - 12115782 - 12151189 on NCBI Build GRCh38
          Downstream ubiquitin specific peptidase 17-like family member 7 USP17L7 3.1kb Sense Chr. 8 - 12132417 - 12134009 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800009635.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000986.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3338491 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4158 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4159 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 3279 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3338491 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3338491 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959798 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16069649 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16069578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16069578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959798 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16069649 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959798 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8094135 - -
          Affymetrix Human Transcriptome Array 2.0 PSR08012377.hg.1 NR_003275.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08012377.hg.1 TC08000986.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0800165513.hg.1 NR_003275.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800165513.hg.1 TC0800009635.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800165513.hg.1 NR_003275.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800165514.hg.1 ENST00000526505.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800165514.hg.1 NR_003275.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800165514.hg.1 TC0800009635.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800165514.hg.1 ENST00000526505.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800165514.hg.1 NR_003275.2 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035530 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035530 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2092496 NR_003275.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2092496 BF947332.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2092496 NR_003275.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2092496 BF947332.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2092496 NR_003275.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2092496 BF947332.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2092496 NR_003275.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2092496 BF947332.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2092496 NR_003275.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2092496 BF947332.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2092496 NR_003275.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2092496 BF947332.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2092496 NR_003275.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2092496 BF947332.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2092496 NR_003275.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2092496 BF947332.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix