Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000497969.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000578195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000579526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
ENST00000582557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012223.hg.1 |
NR_024187.2 |
Refseq |