Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01932692_s1
          See other ZNF770 GE Assays ›
          Gene Symbol:
          ZNF770
          Gene Name:
          zinc finger protein 770
          Gene Aliases:
          PRO1914
          Chromosome Location:
          Chr.15: 34978341 - 34988296 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01932692_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZNF770
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014106.3 NP_054825.2 3 1579 119
          XM_011521744.2 XP_011520046.1 2 1423 119
          GenBank mRNA AK131433.1 - 1 451 119
          AK291438.1 - 3 1570 119
          BC071603.1 - 3 1551 30340520 119
          BM454360.1 - 1 100 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500009005.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001203.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK131433.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ZNF770

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15010794.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15010794.hg.1 NM_014106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15010794.hg.1 TC15001203.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 ENST00000559564.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 ENST00000559564.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 ENST00000559564.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15010792.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15010792.hg.1 NM_014106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15010792.hg.1 TC15001203.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500165147.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165147.hg.1 NM_014106.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500165147.hg.1 TC1500009005.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500165147.hg.1 NM_014106.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500165147.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 ENST00000356321.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 ENST00000559564.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 ENST00000559564.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 ENST00000559564.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 NM_014106.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009005.hg.1 NM_014106.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0