Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01940810_u1
          See other OR4P4 GE Assays ›
          Gene Symbol:
          OR4P4
          Gene Name:
          olfactory receptor family 4 subfamily P member 4
          Gene Aliases:
          OR4P3P
          Chromosome Location:
          Chr.11: 55638358 - 55639296 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01940810_u1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR4P4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004124.2 NP_001004124.1 1 759 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100007600.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000433.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR4P4

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          G protein-coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07806164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215266 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215286 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215297 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215282 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07806190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07806183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215269 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215310 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215284 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16452430 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16452429 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16452422 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07806150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215308 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215267 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215281 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215299 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215278 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07806177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215279 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215274 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07806156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07806184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07806174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215288 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16452419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215302 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28215277 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 133469 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3354836 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0287 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0285 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0286 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 3902 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P32908038 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3354836 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3354836 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07806150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215302 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215282 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215300 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215297 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215279 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07806156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215284 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16452422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215292 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215271 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07806184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16452429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215288 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215269 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07806179 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215299 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07806183 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07806150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215288 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215299 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07806156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215292 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215297 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215282 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215302 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215271 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07806183 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215284 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16452422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215300 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215279 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07806184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07806179 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16452429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28215269 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806172 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806169 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806183 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806150 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806177 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806187 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806151 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806153 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806175 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806186 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806159 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806156 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806167 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806185 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806173 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806184 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806164 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806170 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806162 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806180 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806181 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07806157 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P39361721 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39361721 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806192 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806173 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806211 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806220 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806205 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806159 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806169 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806181 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16452426 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806183 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806203 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806215 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16452429 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806187 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806167 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806164 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806175 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806157 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806172 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806152 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806162 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16452420 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806150 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806223 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806208 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806156 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806217 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16452422 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806177 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806190 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806185 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16452431 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07806170 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P39361721 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001004124_at - -
          Affymetrix Human Gene 1.0 ST Array 7939929 - -
          Affymetrix Human Gene 1.1 ST Array 7939928 - -
          Affymetrix Human Genome U219 Array 11742602_at - -
          Affymetrix Human Genome U219 Array 11742602_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11005490.hg.1 ENST00000314612.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005490.hg.1 NM_001004124.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005490.hg.1 TC11000433.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100007600.hg.1 ENST00000314612.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007600.hg.1 NM_001004124.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100153769.hg.1 ENST00000314612.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153769.hg.1 NM_001004124.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100153769.hg.1 TC1100007600.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100153769.hg.1 NM_001004124.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100153769.hg.1 ENST00000314612.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007600.hg.1 ENST00000314612.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007600.hg.1 NM_001004124.2 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046379 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046379 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1804845 NM_001004124.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1804845 NM_001004124.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1804845 NM_001004124.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1804845 NM_001004124.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1804845 NM_001004124.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_3498-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1804845 NM_001004124.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1804845 NM_001004124.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 2600180 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_3498 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1804845 NM_001004124.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1804845 NM_001004124.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1804845 NM_001004124.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1804845 NM_001004124.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1804845 NM_001004124.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0