Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01941699_s1
          See other RBAKDN GE Assays ›
          Gene Symbol:
          RBAKDN RBAK-RBAKDN
          Gene Name:
          RBAK downstream neighbor (non-protein coding)
          RBAK-RBAKDN readthrough
          Gene Aliases:
          -
          RBAK-LOC389458
          Chromosome Location:
          Chr.7: 5072060 - 5073223 on Build GRCh38
          Chromosome Location:
          Chr.7: 5045821 - 5073223 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01941699_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RBAKDN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015343.2 3 496 69
          GenBank mRNA AA398766.1 - 1 69
          AI015600.1 - 1 69
          BC031661.1 - 3 431 69
          CR745855.1 - 3 389 69
          Gene Symbol: RBAK-RBAKDN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001204513.1 NP_001191442.1 6 775 69
          GenBank mRNA BM684776.1 - 1 69
          BU620638.1 - 1 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25491898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05277200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05277203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05277206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05277199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05277196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05277198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05277207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05277209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05277208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15904549 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3232038 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17178 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60334 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3232038 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3232038 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3232038 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25491869 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277167 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25491872 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25491861 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15904535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277179 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15904536 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25491866 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277183 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15904537 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25491869 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15904535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25491861 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15904537 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25491872 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277179 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277183 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15904536 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25491866 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05277167 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P171143 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P171143 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15904547 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277166 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15904546 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277194 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277172 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15904536 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277200 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277190 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277171 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277175 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277204 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277169 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15904539 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277173 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277186 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277183 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15904544 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277198 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15904548 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15904541 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15904534 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277177 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05277208 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_015343_at - -
          Affymetrix Human Gene 1.0 ST Array 8131298 - -
          Affymetrix Human Gene 1.0 ST Array 8131299 - -
          Affymetrix Human Gene 1.0 ST Array 8131300 - -
          Affymetrix Human Gene 1.1 ST Array 8131296 - -
          Affymetrix HT HG-U133+ PM Array Plate 230439_PM_at - -
          Human Genome U133 Plus 2.0 Array 230439_at - -
          Human Genome U133 Plus 2.0 Array 230439_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230439_at4 - -
          Human Genome U95 Set 46259_PM_r_at - -
          Human Genome U95 Set 46259_r_at - -
          Human Genome U133 Set 230439_PM_at - -
          Human Genome U133 Set 230439_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033904 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033904 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033904 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033904 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 149902 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2054938 NM_001204513.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2054938 BM684776.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2054938 BU620638.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2054938 BC031661.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2054938 CR745855.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1753685 - -
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2054938 NM_001204513.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2054938 BM684776.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2054938 BU620638.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2054938 BC031661.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2054938 CR745855.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1753685 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1753685 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2054938 NM_001204513.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2054938 BM684776.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2054938 BU620638.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2054938 BC031661.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2054938 CR745855.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2054938 NM_001204513.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2054938 BM684776.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2054938 BU620638.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2054938 BC031661.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2054938 CR745855.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2054938 NM_001204513.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2054938 BM684776.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2054938 BU620638.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2054938 BC031661.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2054938 CR745855.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2054938 NM_001204513.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2054938 BM684776.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2054938 BU620638.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2054938 BC031661.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2054938 CR745855.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0