Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02518671_g1
          See other LOC401286 GE Assays ›
          Gene Symbol:
          LOC401286
          Gene Name:
          uncharacterized LOC401286
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 167666840 - 167679223 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02518671_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC401286
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_117091.1 3 4044 75
          GenBank mRNA AK127120.1 - 3 4044 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3409245 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21866 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 57898 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3409238 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3409245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8130806 - -
          Affymetrix Human Gene 1.0 ST Array 8130977 - -
          Affymetrix Human Gene 1.0 ST Array 8130978 - -
          Affymetrix Human Gene 1.0 ST Array 8130979 - -
          Affymetrix Human Gene 1.0 ST Array 8130804 - -
          Affymetrix Human Gene 1.0 ST Array 8130805 - -
          Affymetrix Human Gene 1.1 ST Array 8130803 - -
          Affymetrix Human Gene 1.1 ST Array 8130976 - -
          Human Genome U95 Set 60690_PM_at - -
          Human Genome U95 Set 60690_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3307132 AK123853.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3307132 AK127120.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1784493 AK127120.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1784493 AK127120.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1784493 AK127120.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1784493 AK127120.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1784493 AK127120.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1784493 AK127120.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1784493 AK127120.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1784493 AK127120.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1784493 AK127120.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0