Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02811054_g1
          See other R3HCC1L GE Assays ›
          Gene Symbol:
          R3HCC1L
          Gene Name:
          R3H domain and coiled-coil containing 1 like
          Gene Aliases:
          C10orf28, GIDRP86, GIDRP88, PSORT
          Chromosome Location:
          Chr.10: 98134608 - 98244897 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02811054_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: R3HCC1L
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256619.1 NP_001243548.1 8 - 9 2422 96
          NM_001256620.1 NP_001243549.1 6 - 7 2329 96
          NM_001256621.1 NP_001243550.1 5 - 6 526 96
          NM_014472.4 NP_055287.4 6 - 7 2325 96
          NM_138469.2 NP_612478.2 7 - 8 2422 96
          XM_011539639.1 XP_011537941.1 8 - 9 2679 96
          XM_011539640.1 XP_011537942.1 9 - 10 2484 96
          XM_011539641.1 XP_011537943.1 8 - 9 2436 96
          XM_011539642.1 XP_011537944.1 9 - 10 2776 96
          XM_011539643.1 XP_011537945.1 7 - 8 2363 96
          XM_011539644.1 XP_011537946.1 7 - 8 2367 96
          XM_011539645.2 XP_011537947.1 7 - 8 2610 96
          XM_011539646.1 XP_011537948.1 6 - 7 2242 96
          XM_011539647.1 XP_011537949.1 6 - 7 2244 96
          XM_011539648.1 XP_011537950.1 5 - 6 2117 96
          XM_011539649.1 XP_011537951.1 7 - 8 2637 96
          XM_017016074.1 XP_016871563.1 7 - 8 3575 96
          XM_017016075.1 XP_016871564.1 4 - 5 2075 96
          XM_017016076.1 XP_016871565.1 7 - 8 2369 96
          XM_017016077.1 XP_016871566.1 9 - 10 2532 96
          XM_017016078.1 XP_016871567.1 8 - 9 2454 96
          XM_017016079.1 XP_016871568.1 8 - 9 2468 96
          XM_017016080.1 XP_016871569.1 6 - 7 2374 96
          XM_017016081.1 XP_016871570.1 7 - 8 2456 96
          XM_017016082.1 XP_016871571.1 5 - 6 2248 96
          XM_017016083.1 XP_016871572.1 6 - 7 2264 96
          GenBank mRNA AF525304.1 - 8 - 9 2375 96
          AK058187.1 - 2 - 3 116 96
          AK094479.1 - 3 - 4 836 96
          AK292546.1 - 6 - 7 2324 96
          BC009231.2 - 2 - 3 251 96
          BC047908.1 - 5 - 6 472 96
          BC098175.1 - 3 - 4 269 96
          BC098268.1 - 7 - 8 2363 40026913 96
          BC098359.1 - 6 - 7 2266 40026912 96
          BC099848.1 - 6 - 7 2266 40026916 96

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene R3HCC1L

          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0