Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03008769_g1
          See other UGT2B15 GE Assays ›
          Gene Symbol:
          UGT2B15
          Gene Name:
          UDP glucuronosyltransferase family 2 member B15
          Gene Aliases:
          HLUG4, UDPGT 2B8, UDPGT2B15, UDPGTH3, UGT2B8
          Chromosome Location:
          Chr.4: 68646597 - 68670776 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03008769_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UGT2B15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001076.3 NP_001067.2 1 504 138
          GenBank mRNA AF180322.1 - 1 384 138
          AF548389.1 - 1 363 138
          AK289419.1 - 1 377 138
          BC171879.1 - 504 9054533 138
          U06641.1 - 1 138
          U08854.1 - 1 367 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400012924.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001261.hg.1, TC4_ctg9_hap1000004.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene UGT2B15

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          steroid metabolic process
          flavonoid biosynthetic process
          cellular glucuronidation
          flavonoid glucuronidation
          retinoic acid binding
          glucuronosyltransferase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04017165.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017165.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017165.hg.1 TC04001261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04017168.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017168.hg.1 TC04001261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000057.hg.1 TC4_ctg9_hap1000004.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000057.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000057.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400010907.hg.1 ENST00000509021.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400010907.hg.1 ENST00000509021.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000056.hg.1 TC4_ctg9_hap1000004.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000056.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000056.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017162.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017162.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017162.hg.1 TC04001261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04017166.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017166.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017166.hg.1 TC04001261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000055.hg.1 TC4_ctg9_hap1000004.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000055.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000055.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400010910.hg.1 ENST00000510547.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400010910.hg.1 ENST00000510547.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017163.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017163.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017163.hg.1 TC04001261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000054.hg.1 TC4_ctg9_hap1000004.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000054.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000054.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017164.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017164.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017164.hg.1 TC04001261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0400007751.hg.1 ENST00000507775.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007751.hg.1 ENST00000507775.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007751.hg.1 ENST00000507775.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000058.hg.1 TC4_ctg9_hap1000004.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000058.hg.1 NM_001076.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400010919.hg.1 ENST00000514659.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190172.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190172.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400190172.hg.1 TC0400012924.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190172.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400190172.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400010910.hg.1 ENST00000510547.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400010910.hg.1 ENST00000510547.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190173.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190173.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400190173.hg.1 TC0400012924.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190173.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190173.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400007751.hg.1 ENST00000507775.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400007751.hg.1 ENST00000507775.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400007751.hg.1 ENST00000507775.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400010919.hg.1 ENST00000514659.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190175.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190175.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400190175.hg.1 TC0400012924.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190175.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190175.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400010907.hg.1 ENST00000509021.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400010907.hg.1 ENST00000509021.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190178.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400190178.hg.1 TC0400012924.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190178.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400190174.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190174.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400190174.hg.1 TC0400012924.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190174.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190174.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012924.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 ENST00000317746.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012923.hg.1 NM_001077.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400190176.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190176.hg.1 NM_001076.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400190176.hg.1 TC0400012924.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190176.hg.1 ENST00000338206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190176.hg.1 NM_001076.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 100569 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0