Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03044222_m1
          See other MARVELD1 GE Assays ›
          Gene Symbol:
          MARVELD1
          Gene Name:
          MARVEL domain containing 1
          Gene Aliases:
          GB14, MARVD1, MRVLDC1, bA548K23.8
          Chromosome Location:
          Chr.10: 97713708 - 97718152 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03044222_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MARVELD1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031484.3 NP_113672.1 1 - 2 2273 111
          GenBank mRNA AK025996.1 - 1 - 2 1958 111
          AK027093.1 - 1 - 2 723 111
          BC004995.1 - 1 - 2 2257 111
          BC062544.1 - 1 - 2 2259 111
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX371337.2 - 1 - 2 Chr. 10 - 97715231 - 97717338 [-] on Build GRCh38 111
          GenBankmRNA AI244268.1 - 1 - 2 136 Chr. 10 - 97715845 - 97717482 [+] on Build GRCh38 111
          GenBankmRNA BF983134.1 - 1 - 2 517 Chr. 10 - 97715465 - 97717377 [+] on Build GRCh38 111
          GenBankmRNA BX090352.1 - 1 - 2 416 Chr. 10 - 97715565 - 97717477 [+] on Build GRCh38 111
          GenBankmRNA BG031021.1 - 2 - 3 496 Chr. 10 - 97714694 - 97717493 [+] on Build GRCh38 111
          GenBankmRNA BI858201.1 - 3 - 4 250 Chr. 10 - 97713517 - 97717627 [+] on Build GRCh38 111
          GenBankmRNA CN399819.1 - 1 - 2 294 Chr. 10 - 97715699 - 97717467 [+] on Build GRCh38 111
          GenBankmRNA AW630593.1 - 1 - 2 249 Chr. 10 - 97715732 - 97717345 [+] on Build GRCh38 111
          GenBankmRNA U46299.1 - 1 - 2 116 Chr. 10 - 97715865 - 97717314 [+] on Build GRCh38 111
          GenBankmRNA BE546324.1 - 1 - 2 211 Chr. 10 - 97715769 - 97717737 [+] on Build GRCh38 111
          GenBankmRNA BE273183.1 - 1 - 2 95 Chr. 10 - 97715886 - 97717806 [+] on Build GRCh38 111
          GenBankmRNA BI760667.1 - 590 Chr. - - [-] on Build 111
          GenBankmRNA BI200204.1 - 282 Chr. - - [-] on Build 111
          GenBankmRNA BM564996.1 - 1 - 2 183 Chr. 10 - 97715803 - 97717555 [+] on Build GRCh38 111
          GenBankmRNA BQ722260.1 - 1 - 2 119 Chr. 10 - 97715862 - 97717905 [+] on Build GRCh38 111
          GenBankmRNA W81343.1 - 322 Chr. - - [-] on Build 111
          GenBankmRNA BQ671253.1 - 1 - 2 155 Chr. 10 - 97715826 - 97717683 [+] on Build GRCh38 111
          GenBankmRNA BU789782.1 - 1 - 2 183 Chr. 10 - 97715803 - 97717585 [+] on Build GRCh38 111
          GenBankmRNA BG680011.1 - 1 - 2 126 Chr. 10 - 97715855 - 97718007 [+] on Build GRCh38 111
          GenBankmRNA AW966550.1 - 1 - 2 242 Chr. 10 - 97715739 - 97717553 [+] on Build GRCh38 111
          GenBankmRNA BE784598.1 - 185 Chr. - - [-] on Build 111
          GenBankmRNA BI198003.1 - 218 Chr. - - [-] on Build 111
          GenBankmRNA DA570116.1 - 1 - 2 440 Chr. 10 - 97715541 - 97717495 [+] on Build GRCh38 111
          GenBankmRNA BU146020.1 - 1 - 2 668 Chr. 10 - 97715320 - 97717429 [+] on Build GRCh38 111
          GenBankmRNA BX389381.1 - 1 - 2 145 Chr. 10 - 97715835 - 97717896 [+] on Build GRCh38 111
          GenBankmRNA BU844661.1 - 1 - 2 695 Chr. 10 - 97715286 - 97717387 [+] on Build GRCh38 111
          GenBankmRNA BM702955.1 - 1 - 2 143 Chr. 10 - 97715838 - 97717271 [+] on Build GRCh38 111
          GenBankmRNA BM756030.1 - 1 - 2 141 Chr. 10 - 97715840 - 97717530 [+] on Build GRCh38 111
          GenBankmRNA BQ685699.1 - 1 - 2 566 Chr. 10 - 97715415 - 97717395 [+] on Build GRCh38 111
          GenBankmRNA BG109608.1 - 1 - 2 341 Chr. 10 - 97715640 - 97717687 [+] on Build GRCh38 111
          GenBankmRNA BG421543.1 - 1 - 2 132 Chr. 10 - 97715849 - 97717865 [+] on Build GRCh38 111
          GenBankmRNA BI756017.1 - 1 - 2 431 Chr. 10 - 97715561 - 97717627 [+] on Build GRCh38 111
          GenBankmRNA BI858595.1 - 1 - 2 333 Chr. 10 - 97715647 - 97717730 [+] on Build GRCh38 111
          GenBankmRNA BF316005.1 - 1 - 2 217 Chr. 10 - 97715765 - 97717752 [+] on Build GRCh38 111
          GenBankmRNA BG750034.1 - 1 - 2 383 Chr. 10 - 97715599 - 97717606 [+] on Build GRCh38 111
          GenBankmRNA BM788130.1 - 1 - 2 363 Chr. 10 - 97715618 - 97717514 [+] on Build GRCh38 111
          GenBankmRNA BM739141.1 - 1 - 2 364 Chr. 10 - 97715618 - 97717408 [+] on Build GRCh38 111
          GenBankmRNA BQ688723.1 - 1 - 2 569 Chr. 10 - 97715414 - 97717475 [+] on Build GRCh38 112
          GenBankmRNA BF304030.1 - 1 - 2 491 Chr. 10 - 97715507 - 97717460 [+] on Build GRCh38 111
          GenBankmRNA CN343258.1 - 1 - 2 285 Chr. 10 - 97715696 - 97717551 [+] on Build GRCh38 111

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream arginine vasopressin induced 1 AVPI1 PP5395,
          VIP32,
          VIT32
          -26.5kb Anti-sense Chr. 10 - 97677422 - 97687432 on NCBI Build GRCh38
          Downstream zinc finger FYVE-type containing 27 ZFYVE27 PROTRUDIN,
          SPG33
          19.0kb Sense Chr. 10 - 97737121 - 97760907 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MARVELD1

          Gene Ontology Categories:

          Function(s)

          cell cycle

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix