Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03297669_s1
          See other INGX GE Assays ›
          Gene Symbol:
          INGX TAF1
          Gene Name:
          inhibitor of growth family, X-linked (pseudogene)
          TATA-box binding protein associated factor 1
          Gene Aliases:
          ING1-like, ING2
          BA2R, CCG1, CCGS, DYT3, DYT3/TAF1, KAT4, MRXS33, N-TAF1, NSCL2, OF, P250, TAF(II)250, TAF2A, TAFII-250, TAFII250, XDP
          Chromosome Location:
          Chr.X: 71491681 - 71492449 on Build GRCh38
          Chromosome Location:
          Chr.X: 71366220 - 71530525 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03297669_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: INGX
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002226.3 1 352 69
          GenBank mRNA AF149724.1 - 1 353 69
          BC101124.1 - 1 272 69
          Gene Symbol: TAF1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104396.1 1 69
          GenBank mRNA AJ549246.2 - 1 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream integrin subunit beta 1 binding protein 2 ITGB1BP2 CHORDC3,
          ITGB1BP,
          MELUSIN,
          MSTP015
          -186.3kb Anti-sense Chr. X - 71301275 - 71305371 on NCBI Build GRCh38
          Downstream O-linked N-acetylglucosamine (GlcNAc) transferase OGT HINCUT-1,
          HRNT1,
          O-GLCNAC
          40.6kb Anti-sense Chr. X - 71533062 - 71575897 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00007568.hg.1, TC0X00010002.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X001127.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AJ549246.2 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA-templated transcription, initiation
          transcription by RNA polymerase II
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          cellular response to DNA damage stimulus
          cell cycle
          viral process
          histone acetylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of transcription by RNA polymerase II
          protein autophosphorylation
          RNA polymerase II preinitiation complex assembly
          positive regulation of transcription initiation from RNA polymerase II promoter
          regulation of signal transduction by p53 class mediator
          transcription cis-regulatory region binding
          p53 binding
          transcription coactivator activity
          histone acetyltransferase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          transcription factor binding
          TBP-class protein binding
          sequence-specific DNA binding
          lysine-acetylated histone binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17355701 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11784181 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 59967 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 29693 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 131444 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P125686 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P125686 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43644 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 26071 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P125686 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P125686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11784180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17355701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32499215 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11784183 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32499216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11784181 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11784185 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11784185 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11784180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17355701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11784181 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32499215 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11784183 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32499216 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P125686 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P125686 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17355701 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11784183 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11784181 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17355704 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11784185 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11784174 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11784180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11784175 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17355699 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17355700 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P121425 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002226_at - -
          Affymetrix Human Gene 1.0 ST Array 8173487 - -
          Affymetrix Human Gene 1.0 ST Array 8173491 - -
          Affymetrix Human Gene 1.0 ST Array 8173488 - -
          Affymetrix Human Gene 1.0 ST Array 8173490 - -
          Affymetrix Human Gene 1.0 ST Array 8173489 - -
          Affymetrix Human Gene 1.0 ST Array 8173492 - -
          Affymetrix Human Gene 1.1 ST Array 8173486 - -
          Affymetrix HT HG-U133+ PM Array Plate 224245_PM_at - -
          Affymetrix Human Genome U219 Array 11750371_PM_x_at - -
          Affymetrix Human Genome U219 Array 11757250_PM_x_at - -
          Affymetrix Human Genome U219 Array 11763946_PM_at - -
          Affymetrix Human Genome U219 Array 11757250_x_at - -
          Affymetrix Human Genome U219 Array 11757249_PM_at - -
          Affymetrix Human Genome U219 Array 11750371_x_at - -
          Affymetrix Human Genome U219 Array 11763946_at - -
          Affymetrix Human Genome U219 Array 11757249_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR0X015352.hg.1 NR_002226.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X015352.hg.1 TC0X001127.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0X00010002.hg.1 ENST00000359239.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010002.hg.1 ENST00000359239.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010002.hg.1 ENST00000359239.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010002.hg.1 ENST00000489074.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010002.hg.1 ENST00000489074.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010002.hg.1 NR_002226.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010002.hg.1 NR_002226.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00152098.hg.1 NR_104396.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00152098.hg.1 TC0X00007568.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00152098.hg.1 NR_104396.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00168283.hg.1 ENST00000489074.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00168283.hg.1 NR_002226.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00168283.hg.1 TC0X00010002.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00168283.hg.1 NR_002226.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00168283.hg.1 ENST00000489074.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010002.hg.1 ENST00000359239.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010002.hg.1 ENST00000359239.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010002.hg.1 ENST00000359239.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010002.hg.1 ENST00000489074.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010002.hg.1 ENST00000489074.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010002.hg.1 NR_002226.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00010002.hg.1 NR_002226.3 Refseq
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_002226_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040041 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040041 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 134829 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2196287 BC101124.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2196287 BC117297.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2196287 NR_002226.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2196287 AF149724.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1783274 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2196287 BC101124.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2196287 BC117297.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2196287 NR_002226.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2196287 AF149724.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2196287 BC101124.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2196287 BC117297.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2196287 NR_002226.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2196287 AF149724.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2196287 BC101124.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2196287 BC117297.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2196287 NR_002226.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2196287 AF149724.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1783274 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 940390 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_25409 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2196287 BC101124.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2196287 BC117297.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2196287 NR_002226.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2196287 AF149724.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2196287 BC101124.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2196287 BC117297.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2196287 NR_002226.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2196287 AF149724.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2196287 BC101124.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2196287 BC117297.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2196287 NR_002226.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2196287 AF149724.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1783274 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2196287 BC101124.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2196287 BC117297.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2196287 NR_002226.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2196287 AF149724.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0