Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03297859_s1
          See other SNORA33 GE Assays ›
          Gene Symbol:
          SNORA33
          Gene Name:
          small nucleolar RNA, H/ACA box 33
          Gene Aliases:
          ACA33
          Chromosome Location:
          Chr.6: 132817219 - 132817351 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03297859_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORA33
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002436.1 1 61 80
          GenBank mRNA AJ609473.1 - 1 61 80
          AK096147.1 - 1 2708 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW272582.1 - 1 Chr. 6 - 132817186 - 132817564 [-] on Build GRCh38 80
          GenBankmRNA AI887274.1 - 1 Chr. 6 - 132817032 - 132817564 [-] on Build GRCh38 80
          GenBankmRNA DW441862.1 - 1 77 Chr. 6 - 132817219 - 132817439 [+] on Build GRCh38 80
          GenBankmRNA DB321463.1 - 1 Chr. 6 - 132817111 - 132817567 [-] on Build GRCh38 80
          GenBankmRNA AW103472.1 - 1 Chr. 6 - 132816968 - 132817567 [-] on Build GRCh38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          C/D box 100
          SNORD100 HBII-429 -342.0 Sense Chr. 6 - 132816802 - 132816877 on NCBI Build GRCh38
          Downstream long intergenic non-protein coding RNA 326 LINC00326 NCRNA00326 270.7kb Sense Chr. 6 - 133088080 - 133106578 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600009491.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000997.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3770 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25285787 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25285786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25285786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25285787 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05085510 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05085511 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002436_at - -
          Affymetrix Human Gene 1.0 ST Array 8122143 - -
          Affymetrix Human Gene 1.0 ST Array 8122141 - -
          Affymetrix Human Gene 1.0 ST Array 8122145 - -
          Affymetrix Human Gene 1.0 ST Array 8122140 - -
          Affymetrix Human Gene 1.0 ST Array 8122139 - -
          Affymetrix Human Gene 1.1 ST Array 8122136 - -
          Affymetrix Human Gene 1.1 ST Array 8122144 - -
          Affymetrix Human Gene 1.1 ST Array 8122142 - -
          Affymetrix Human Genome U219 Array 11763888_at - -
          Affymetrix Human Genome U219 Array 11763888_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009489.hg.1 ENST00000384027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009489.hg.1 NR_002434.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009490.hg.1 ENST00000408573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009490.hg.1 NR_002435.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009491.hg.1 ENST00000363664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009491.hg.1 NR_002436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011748.hg.1 ENST00000363664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011748.hg.1 NR_002436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011748.hg.1 TC06000997.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009489.hg.1 ENST00000384027.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009489.hg.1 NR_002434.1 Refseq
          Affymetrix Human Clariom D Assay TC0600009490.hg.1 ENST00000408573.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009490.hg.1 NR_002435.1 Refseq
          Affymetrix Human Clariom D Assay TC0600009491.hg.1 ENST00000363664.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009491.hg.1 NR_002436.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165081.hg.1 ENST00000363664.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165081.hg.1 NR_002436.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165081.hg.1 TC0600009491.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600165081.hg.1 NR_002436.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165081.hg.1 ENST00000363664.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 143873 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2096747 AK096147.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2096747 AJ609473.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2096747 NR_002436.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1705715 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2096747 AK096147.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2096747 AJ609473.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2096747 NR_002436.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2096747 AK096147.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2096747 AJ609473.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2096747 NR_002436.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2096747 AK096147.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2096747 AJ609473.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2096747 NR_002436.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1705715 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 4280491 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_10854 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2096747 AK096147.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2096747 AJ609473.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2096747 NR_002436.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2096747 AK096147.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2096747 AJ609473.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2096747 NR_002436.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1705715 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix