Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298241_s1
          See other TSPY26P GE Assays ›
          Gene Symbol:
          TSPY26P
          Gene Name:
          testis specific protein, Y-linked 26, pseudogene
          Gene Aliases:
          TSPYL3, bA392M18.1
          Chromosome Location:
          Chr.20: 32189146 - 32190360 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03298241_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TSPY26P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002781.1 1 956 123
          GenBank mRNA BC101556.1 - 1 956 123
          BC101558.1 - 1 956 123
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BQ432315.1 - 1 479 Chr. 20 - 32189136 - 32189883 [-] on Build GRCh38 123
          GenBankmRNA CN430810.1 - 1 113 Chr. 20 - 32188777 - 32189517 [-] on Build GRCh38 123
          GenBankmRNA AL935106.1 - 1 151 Chr. 20 - 32188836 - 32189554 [-] on Build GRCh38 123

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transmembrane 9 superfamily member 4 TM9SF4 dJ836N17.2 -21.9kb Anti-sense Chr. 20 - 32109506 - 32167258 on NCBI Build GRCh38
          Downstream PLAG1 like zinc finger 2 PLAGL2 ZNF900 2.1kb Sense Chr. 20 - 32192504 - 32207743 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000008844.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s)

          nucleosome assembly

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P408341 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P408341 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 4468 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 34298 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P408341 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P408341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132998 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132991 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31780149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31780178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31780174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31780156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133010 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133000 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132993 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132998 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133000 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133010 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31780149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31780178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31780174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132991 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132993 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11132994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31780156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11133015 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P5994 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P408341 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P102816 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P408341 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P34683926 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132993 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17186829 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132968 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133017 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133012 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132988 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133040 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132970 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132960 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133002 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132964 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132972 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133003 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132974 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132984 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133007 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132978 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133020 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132998 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133004 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132981 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132952 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132987 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132997 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133000 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133015 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132961 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132953 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132957 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133010 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132991 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11132996 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11133019 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002781_at - -
          Affymetrix Human Gene 1.0 ST Array 8065605 - -
          Affymetrix Human Gene 1.0 ST Array 8065606 - -
          Affymetrix Human Gene 1.0 ST Array 8065604 - -
          Affymetrix Human Gene 1.1 ST Array 8065603 - -
          Affymetrix HT HG-U133+ PM Array Plate 233617_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2000008844.hg.1 ENST00000476365.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008844.hg.1 ENST00000476365.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008844.hg.1 ENST00000565928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008844.hg.1 ENST00000565928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008844.hg.1 ENST00000565928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008844.hg.1 ENST00000565928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008844.hg.1 NR_002781.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008844.hg.1 NR_002781.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008844.hg.1 ENST00000476365.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008844.hg.1 ENST00000476365.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008844.hg.1 ENST00000565928.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008844.hg.1 ENST00000565928.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008844.hg.1 ENST00000565928.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008844.hg.1 ENST00000565928.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008844.hg.1 NR_002781.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008844.hg.1 NR_002781.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000161663.hg.1 ENST00000565928.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000161663.hg.1 NR_002781.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000161663.hg.1 TC2000008844.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000161663.hg.1 NR_002781.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000161663.hg.1 ENST00000565928.1 Ensembl
          Human Genome U133 Plus 2.0 Array 233617_at - -
          Human Genome U133 Plus 2.0 Array 233617_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_002781_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233617_at1 - -
          Human Genome U133 Set 233617_at - -
          Human Genome U133 Set 233617_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A017550 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A017550 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2231478 BC101556.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2231478 BC101558.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2231478 NR_002781.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1737360 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2231478 BC101556.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2231478 BC101558.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2231478 NR_002781.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2231478 BC101556.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2231478 BC101558.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2231478 NR_002781.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2231478 BC101556.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2231478 BC101558.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2231478 NR_002781.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1737360 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2231478 BC101556.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2231478 BC101558.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2231478 NR_002781.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2231478 BC101556.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2231478 BC101558.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2231478 NR_002781.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1737360 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2231478 BC101556.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2231478 BC101558.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2231478 NR_002781.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2231478 BC101556.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2231478 BC101558.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2231478 NR_002781.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0