Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298347_s1
          See other LOC256880 GE Assays ›
          Gene Symbol:
          LOC256880
          Gene Name:
          uncharacterized LOC256880
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 99950494 - 100037705 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03298347_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC256880
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK123926.1 - 1 1650 120
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY237283.1 - 1 Chr. 4 - 99952029 - 99952463 [-] on Build GRCh38 120
          GenBankmRNA DB327895.1 - 1 Chr. 4 - 99951929 - 99952463 [-] on Build GRCh38 120

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream H2A histone family member Z H2AFZ H2A.Z-1,
          H2A.z,
          H2A/z,
          H2AZ
          -1.6kb Sense Chr. 4 - 99948087 - 99950355 on NCBI Build GRCh38
          Downstream DNA damage inducible transcript 4 like DDIT4L REDD2,
          Rtp801L
          232.6kb Sense Chr. 4 - 100185870 - 100190498 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3294583 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3886 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3887 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 16983 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3294583 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3294583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459757 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15596121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459739 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15596116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15596123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03300830 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03300835 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459768 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459736 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03300816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15596115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03300809 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459770 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459736 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459768 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03300816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15596115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03300835 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03300809 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459757 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03300830 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15596116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15596123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15596121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459739 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459770 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23459751 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300824 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300849 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300812 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300828 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15596118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300818 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300833 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300842 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300805 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300844 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300851 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300838 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300826 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300793 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15596120 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15596124 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300839 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300815 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300830 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300807 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300847 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300811 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15596116 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15596119 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15596123 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300831 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300835 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03300821 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025834 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025834 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3246909 NR_002799.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3246909 AK123926.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3246909 NR_002799.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3246909 AK123926.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3246909 NR_002799.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3246909 AK123926.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3246909 NR_002799.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3246909 AK123926.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0