Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298665_m1
          See other GAS2L1P2 GE Assays ›
          Gene Symbol:
          GAS2L1P2
          Gene Name:
          growth arrest specific 2 like 1 pseudogene 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 97075671 - 97081945 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03298665_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GAS2L1P2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002942.1 2 - 3 241 91
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX436450.2 - 3 - 4 301 Chr. 9 - 97077159 - 97089410 [-] on Build GRCh38 91
          GenBankmRNA BX404824.2 - 3 - 4 301 Chr. 9 - 97076885 - 97089410 [-] on Build GRCh38 91
          GenBankmRNA BI831266.1 - 2 - 3 242 Chr. 9 - 97077304 - 97081945 [-] on Build GRCh38 91
          GenBankmRNA HY071092.1 - 3 - 4 318 Chr. 9 - 97077610 - 97089427 [-] on Build GRCh38 91
          GenBankmRNA BX447324.2 - 3 - 4 301 Chr. 9 - 97076877 - 97089410 [-] on Build GRCh38 91

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cathepsin V CTSV CATL2,
          CTSL2,
          CTSU
          -36.0kb Sense Chr. 9 - 97029677 - 97039643 on NCBI Build GRCh38
          Downstream uncharacterized LOC105376166 LOC105376166 48.8kb Sense Chr. 9 - 97123933 - 97130679 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900012259.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001382.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16245400 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16245402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836790 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 860104 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 385816 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 156903 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 325207 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P753476 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P753476 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 11344 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2875 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P753476 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P753476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27179495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27179505 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836787 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16245374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27179496 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836787 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27179495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16245374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27179505 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27179496 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P753476 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P753476 - -
          Agilent Human 1B Microarray G4111A A_32_P758899 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836791 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836781 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836789 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836775 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836782 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16245362 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836778 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836785 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836786 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836787 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16245353 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 ENST00000436355.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 ENST00000436355.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010054.hg.1 ENST00000420145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017298.hg.1 ENST00000442849.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017298.hg.1 NR_002942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017298.hg.1 TC09001382.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09017295.hg.1 NR_002942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017295.hg.1 TC09001382.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09017297.hg.1 ENST00000442849.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017297.hg.1 NR_002942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017297.hg.1 TC09001382.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09017296.hg.1 ENST00000442849.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017296.hg.1 NR_002942.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017296.hg.1 TC09001382.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900186372.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186372.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900186372.hg.1 TC0900012259.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900186372.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900186372.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186369.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186369.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900186369.hg.1 TC0900012259.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900186369.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186369.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900186370.hg.1 ENST00000436355.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186370.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186370.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900186370.hg.1 TC0900012259.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900186370.hg.1 ENST00000436355.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186370.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186370.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 ENST00000436355.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 ENST00000436355.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay TC0900012259.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900186374.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186374.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900186374.hg.1 TC0900012259.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900186374.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186374.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900186371.hg.1 ENST00000436355.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186371.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186371.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900186371.hg.1 TC0900012259.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900186371.hg.1 ENST00000436355.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186371.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186371.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010054.hg.1 ENST00000420145.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186375.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186375.hg.1 NR_002942.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900186375.hg.1 TC0900012259.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900186375.hg.1 ENST00000442849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186375.hg.1 NR_002942.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037775 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037776 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037775 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037776 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3248566 NR_002942.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3248566 BC043374.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3248566 NR_002942.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3248566 BC043374.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3248566 NR_002942.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3248566 BC043374.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3248566 NR_002942.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3248566 BC043374.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix