Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298715_s1
          See other SCARNA6 GE Assays ›
          Gene Symbol:
          SCARNA6
          Gene Name:
          small Cajal body-specific RNA 6
          Gene Aliases:
          U88
          Chromosome Location:
          Chr.2: 233288676 - 233288941 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03298715_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SCARNA6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003006.1 1 48 103
          GenBank mRNA AY077738.1 - 1 48 103

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small Cajal body-specific RNA 5 SCARNA5 U87 -12.7kb Sense Chr. 2 - 233275726 - 233276003 on NCBI Build GRCh38
          Downstream S-antigen visual arrestin SAG RP47,
          S-AG
          15.4kb Sense Chr. 2 - 233307663 - 233347066 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200011127.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001417.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3230837 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4027 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4026 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49793 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3230837 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3230837 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093893 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966599 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966592 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966589 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966617 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15394144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093915 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966613 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093862 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966582 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966595 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966593 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966608 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966601 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966584 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093857 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966603 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093887 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093904 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093883 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093872 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966590 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966598 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15394148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966561 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093903 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093856 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966608 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966613 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093872 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966593 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966561 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966590 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093903 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966603 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093904 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966582 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966601 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966584 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15394144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093915 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966589 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966598 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15394148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093883 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093887 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093893 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093857 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093862 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093856 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966592 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966595 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966599 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22093858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01966617 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966610 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966586 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966588 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966606 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966573 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966603 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966574 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966611 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15394148 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966593 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15394144 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966567 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966555 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966568 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966564 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966582 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966580 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966590 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966608 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966570 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966551 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966705 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15394146 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966599 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966554 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966703 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966601 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966612 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966595 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966563 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966560 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966614 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966558 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966605 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01966578 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_003006_at - -
          Affymetrix Human Gene 1.0 ST Array 8049300 - -
          Affymetrix Human Gene 1.1 ST Array 8049299 - -
          Affymetrix Human Transcriptome Array 2.0 TC0200011127.hg.1 ENST00000515982.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011127.hg.1 NR_003006.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02022564.hg.1 ENST00000515982.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02022564.hg.1 NR_003006.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02022564.hg.1 TC02001417.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0200011127.hg.1 ENST00000515982.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011127.hg.1 NR_003006.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200180237.hg.1 ENST00000515982.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200180237.hg.1 NR_003006.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200180237.hg.1 TC0200011127.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200180237.hg.1 NR_003006.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200180237.hg.1 ENST00000515982.1 Ensembl
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A017115 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A017115 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3241869 NR_003006.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3241869 AY077738.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3241869 NR_003006.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3241869 AY077738.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3241869 NR_003006.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3241869 AY077738.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3241869 NR_003006.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3241869 AY077738.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0