Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03300798_m1
          See other SCAND2P GE Assays ›
          Gene Symbol:
          SCAND2P
          Gene Name:
          SCAN domain containing 2 pseudogene
          Gene Aliases:
          SCAND2
          Chromosome Location:
          Chr.15: 84631460 - 84642463 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03300798_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SCAND2P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003654.2 2 - 3 912 70
          NR_004859.1 3 - 4 1470 70
          GenBank mRNA AA280726.1 - 2 - 3 174 70
          AF229246.1 - 3 - 4 2730 70
          AI218957.1 - 2 - 3 174 70
          AK022844.1 - 2 - 3 912 70
          AK290489.1 - 2 - 3 922 70
          AL109690.1 - 2 - 3 1470 70
          AW341587.1 - 3 - 4 254 70
          BC011547.1 - 2 - 3 894 70
          BC012929.1 - 2 - 3 894 70
          BC063836.1 - 2 - 3 868 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX370292.2 - 2 - 3 192 Chr. 15 - 84632184 - 84640112 [+] on Build GRCh38 70
          GenBankmRNA HY133695.1 - 2 - 3 200 Chr. 15 - 84634256 - 84639690 [+] on Build GRCh38 70
          GenBankmRNA AL357213.1 - 3 - 4 Chr. 15 - 84634230 - 84645664 [-] on Build GRCh38 70
          GenBankmRNA AW085215.1 - 2 - 3 174 Chr. 15 - 84634281 - 84641210 [+] on Build GRCh38 70
          GenBankmRNA BG756069.1 - 2 - 3 161 Chr. 15 - 84634295 - 84640080 [+] on Build GRCh38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger and SCAN domain containing 2 ZSCAN2 ZFP29,
          ZNF854
          -13.6kb Sense Chr. 15 - 84600986 - 84623716 on NCBI Build GRCh38
          Downstream WD repeat domain 73 WDR73 GAMOS,
          HSPC264
          3.0kb Anti-sense Chr. 15 - 84640682 - 84654343 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500008156.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          biological_process
          molecular_function
          DNA-binding transcription factor activity
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000348993.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000530891.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000558508.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000560416.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000560543.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000560678.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 NR_003654.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 NR_004859.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 TC1500008156.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000348993.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000530891.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000558508.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000560416.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000560543.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 ENST00000560678.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 NR_003654.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500158711.hg.1 NR_004859.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0