Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03301058_s1
          See other SNORD11B GE Assays ›
          Gene Symbol:
          SNORD11B
          Gene Name:
          small nucleolar RNA, C/D box 11B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 202291317 - 202291428 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03301058_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORD11B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003694.1 1 60 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          C/D box 70
          SNORD70 HBII-234,
          SNORD70A
          -14.8kb Sense Chr. 2 - 202276431 - 202276518 on NCBI Build GRCh38
          Downstream small nucleolar RNA,
          C/D box 11
          SNORD11 HBII-95 1.6kb Sense Chr. 2 - 202293051 - 202293134 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200010482.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001190.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818564 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943411 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818562 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943419 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818563 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818565 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943433 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818556 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943418 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943428 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818555 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818569 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943409 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943411 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943428 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943433 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943419 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943409 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818565 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943418 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818556 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818564 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818563 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818555 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818562 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01818569 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21943427 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818574 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818582 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818585 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818591 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818562 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818577 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15373674 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818576 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818558 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818589 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818553 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818552 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818565 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818568 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818560 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818572 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818579 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818561 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01818584 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200010482.hg.1 ENST00000607707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010482.hg.1 NR_003694.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018118.hg.1 ENST00000459250.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018118.hg.1 NR_003694.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018118.hg.1 TC02001190.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200174175.hg.1 ENST00000607707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174175.hg.1 NR_003694.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200174175.hg.1 TC0200010482.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200174175.hg.1 NR_003694.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200174175.hg.1 ENST00000607707.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010482.hg.1 ENST00000607707.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010482.hg.1 NR_003694.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3238478 NR_003694.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3238478 NR_003694.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3238478 NR_003694.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3238478 NR_003694.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix