Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03301933_s1
          See other SNORA14A GE Assays ›
          Gene Symbol:
          SNORA14A
          Gene Name:
          small nucleolar RNA, H/ACA box 14A
          Gene Aliases:
          ACA14a
          Chromosome Location:
          Chr.7: 75943783 - 75943916 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03301933_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORA14A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002955.1 1 73 86
          GenBank mRNA AJ609456.1 - 1 73 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 4651 MIR4651 mir-4651 -28.5kb Sense Chr. 7 - 75915197 - 75915269 on NCBI Build GRCh38
          Downstream transmembrane protein 120A TMEM120A NET29,
          TMPIT
          42.9kb Anti-sense Chr. 7 - 75986837 - 75994674 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700008077.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000478.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15975548 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785920 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785923 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785930 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785913 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15975546 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538106 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538092 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15975546 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15975548 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538106 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785930 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785923 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785913 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538092 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785920 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05538095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25785925 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05538100 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05538097 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15975546 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05538095 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05538093 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05538101 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15975548 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05538104 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05538103 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002955_at - -
          Affymetrix Human Gene 1.0 ST Array 8133689 - -
          Affymetrix Human Gene 1.1 ST Array 8133688 - -
          Affymetrix Human Transcriptome Array 2.0 TC0700008077.hg.1 ENST00000364773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008077.hg.1 NR_002955.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017817.hg.1 ENST00000384452.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017817.hg.1 NR_002956.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07006698.hg.1 ENST00000364773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07006698.hg.1 NR_002955.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07006698.hg.1 TC07000478.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700155983.hg.1 ENST00000364773.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700155983.hg.1 NR_002955.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700155983.hg.1 TC0700008077.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700155983.hg.1 NR_002955.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700155983.hg.1 ENST00000364773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008077.hg.1 ENST00000364773.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008077.hg.1 NR_002955.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017817.hg.1 ENST00000384452.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017817.hg.1 NR_002956.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3241036 NR_002955.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3241036 AJ609456.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3241036 NR_002955.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3241036 AJ609456.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3241036 NR_002955.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3241036 AJ609456.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3241036 NR_002955.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3241036 AJ609456.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0