Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03309489_s1
          See other SNORA36B GE Assays ›
          Gene Symbol:
          SNORA36B
          Gene Name:
          small nucleolar RNA, H/ACA box 36B
          Gene Aliases:
          ACA36b
          Chromosome Location:
          Chr.1: 220200546 - 220200676 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03309489_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORA36B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002994.2 1 67 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 664a MIR664A MIR664,
          MIRN664,
          hsa-mir-664,
          hsa-mir-664a,
          mir-664a
          Sense Chr. 1 - 220200538 - 220200619 on NCBI Build GRCh38
          Downstream aurora kinase A pseudogene 1 AURKAPS1 AurAps1,
          STK6P
          65.5kb Sense Chr. 1 - 220266179 - 220267715 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100017375.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003839.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817713 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880153 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817690 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880135 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15176373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817694 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15176371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880120 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817690 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880120 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15176373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817694 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880135 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880153 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15176371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20880161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817713 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817717 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817698 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817703 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817710 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817712 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817707 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15176376 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817695 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817721 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817719 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15176373 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817723 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817729 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15176368 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817738 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817693 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15176379 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817733 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817701 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817731 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817688 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15176380 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817736 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817727 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15176375 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817740 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817684 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817686 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817690 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00817708 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01058698.hg.1 ENST00000410438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01058698.hg.1 NR_002994.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058698.hg.1 TC01003839.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01058697.hg.1 ENST00000410438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01058697.hg.1 NR_002994.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058697.hg.1 NR_031705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058697.hg.1 TC01003839.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100017375.hg.1 ENST00000410438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017375.hg.1 ENST00000410438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017375.hg.1 ENST00000626086.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017375.hg.1 NR_002994.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017375.hg.1 NR_002994.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017375.hg.1 NR_031705.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100231535.hg.1 ENST00000410438.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100231535.hg.1 ENST00000626086.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100231535.hg.1 NR_002994.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100231535.hg.1 NR_031705.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100231535.hg.1 TC0100017375.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100231535.hg.1 NR_031705.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100231535.hg.1 ENST00000410438.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100231535.hg.1 ENST00000626086.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100231535.hg.1 NR_002994.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100231536.hg.1 ENST00000410438.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100231536.hg.1 NR_002994.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100231536.hg.1 TC0100017375.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100231536.hg.1 ENST00000410438.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100231536.hg.1 NR_002994.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017375.hg.1 ENST00000410438.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017375.hg.1 ENST00000410438.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017375.hg.1 ENST00000626086.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017375.hg.1 NR_002994.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017375.hg.1 NR_002994.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017375.hg.1 NR_031705.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3247494 NR_002994.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3247494 NR_002994.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3247494 NR_002994.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3247494 NR_002994.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0