Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03309517_s1
          See other SNORD93 GE Assays ›
          Gene Symbol:
          SNORD93
          Gene Name:
          small nucleolar RNA, C/D box 93
          Gene Aliases:
          HBII-336
          Chromosome Location:
          Chr.7: 22856613 - 22856686 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03309517_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORD93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003075.1 1 37 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream translocase of outer mitochondrial membrane 7 TOMM7 TOM7 -33.8kb Anti-sense Chr. 7 - 22812632 - 22822852 on NCBI Build GRCh38
          Downstream family with sequence similarity 126 member A FAM126A DRCTNNB1A,
          HCC,
          HLD5,
          HYCC1
          39.2kb Anti-sense Chr. 7 - 22895848 - 23014195 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700006899.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000139.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15917257 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358061 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574951 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574920 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574903 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358076 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574954 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574904 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15917243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574904 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574951 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574903 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358076 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574920 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15917257 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15917243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358061 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05358057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25574954 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358098 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15917257 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358101 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358080 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358059 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358090 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358042 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358065 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358082 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358063 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358056 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358053 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358048 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358061 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358078 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358134 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358108 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15917252 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358131 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358074 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358057 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358123 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358113 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358076 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15917246 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358071 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358085 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358096 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358116 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358070 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358050 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358067 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358092 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358069 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358094 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358054 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358088 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15917243 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358112 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358120 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358114 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358103 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358084 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358125 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358129 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358106 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358062 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05358046 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700006899.hg.1 ENST00000408813.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006899.hg.1 NR_003075.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07002145.hg.1 ENST00000408813.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07002145.hg.1 NR_003075.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07002145.hg.1 TC07000139.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0700006899.hg.1 ENST00000408813.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006899.hg.1 NR_003075.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147698.hg.1 ENST00000408813.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147698.hg.1 NR_003075.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147698.hg.1 TC0700006899.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700147698.hg.1 NR_003075.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147698.hg.1 ENST00000408813.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3246529 NR_003075.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3246529 NR_003075.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3246529 NR_003075.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3246529 NR_003075.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix