Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03309787_s1
          See other KATNBL1P6 GE Assays ›
          Gene Symbol:
          KATNBL1P6
          Gene Name:
          katanin regulatory subunit B1 like 1 pseudogene 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 146801669 - 146803824 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03309787_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KATNBL1P6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003954.1 1 2046 136
          GenBank mRNA BC067801.1 - 1 2046 136

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC101928661 LOC101928661 -202.7kb Sense Chr. 6 - 146594516 - 146598931 on NCBI Build GRCh38
          Downstream STXBP5 antisense RNA 1 STXBP5-AS1 37.6kb Sense Chr. 6 - 146841388 - 147204614 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3214061 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4292 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4293 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43343 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3214061 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3214061 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350895 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350926 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05150657 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05150659 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05150663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15870807 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350902 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15870811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05150660 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15870832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15870828 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350919 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05150652 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350917 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350917 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15870828 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350902 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15870832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350926 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350895 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15870811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05150663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05150659 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05150657 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05150652 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350919 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15870807 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05150660 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25350921 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05150662 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05150656 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05150658 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05150665 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05150653 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05150667 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05150674 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05150654 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05150671 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05150659 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7987182 - -
          Affymetrix Human Gene 1.0 ST Array 8130072 - -
          Affymetrix Human Gene 1.0 ST Array 7987185 - -
          Affymetrix Human Gene 1.0 ST Array 7987188 - -
          Affymetrix Human Gene 1.0 ST Array 7987189 - -
          Affymetrix Human Gene 1.0 ST Array 7987183 - -
          Affymetrix Human Gene 1.0 ST Array 7987184 - -
          Affymetrix Human Gene 1.0 ST Array 7987190 - -
          Affymetrix Human Gene 1.0 ST Array 7987186 - -
          Affymetrix Human Gene 1.0 ST Array 7987187 - -
          Affymetrix Human Gene 1.1 ST Array 8130071 - -
          Affymetrix Human Gene 1.1 ST Array 7987180 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030666 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030665 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030666 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030665 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 157096 - -
          ABI Human Genome Survey Microarray Version 2.0 236438 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3243233 NR_003954.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3243233 BC067801.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3243233 NR_003954.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3243233 BC067801.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3243233 NR_003954.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3243233 BC067801.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3243233 NR_003954.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3243233 BC067801.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0