Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006545.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006545.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006545.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145700.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100145700.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145700.hg.1 |
TC1100006545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100145700.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100145700.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006545.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006545.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006545.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006545.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145697.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145697.hg.1 |
TC1100006545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100145697.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145698.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145698.hg.1 |
TC1100006545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100145698.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145702.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145702.hg.1 |
TC1100006545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100145702.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145699.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100145699.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145699.hg.1 |
TC1100006545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100145699.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100145699.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145701.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100145701.hg.1 |
NR_120534.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145701.hg.1 |
TC1100006545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100145701.hg.1 |
ENST00000455671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100145701.hg.1 |
NR_120534.1 |
Refseq |