Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000564594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000564594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000598619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
ENST00000598619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003292.hg.1 |
ENST00000409454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003292.hg.1 |
ENST00000495658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003292.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003292.hg.1 |
TC15002763.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003289.hg.1 |
ENST00000409454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003289.hg.1 |
ENST00000495658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003289.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003289.hg.1 |
TC15002763.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003291.hg.1 |
ENST00000409454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003291.hg.1 |
ENST00000495658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003291.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003291.hg.1 |
TC15002763.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
TC1500010735.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182233.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000564594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000564594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000598619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
ENST00000598619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010735.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
NR_022014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
TC1500010735.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
ENST00000313559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
ENST00000396644.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
ENST00000396645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
ENST00000409454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
ENST00000495658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182235.hg.1 |
NR_022014.1 |
Refseq |