Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03655582_s1
          See other LOC729603 GE Assays ›
          Gene Symbol:
          LOC729603
          Gene Name:
          calcineurin like EF-hand protein 1 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 160093082 - 160096212 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03655582_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC729603
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003288.2 1 2409 82
          GenBank mRNA AK096733.1 - 1 1454 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream antisense of IGF2R non-protein coding RNA AIRN AIR,
          IGF2R-AS,
          IGF2R-AS1,
          IGF2RAS,
          NCRNA00088
          -86.4kb Anti-sense Chr. 6 - 160003291 - 160007664 on NCBI Build GRCh38
          Downstream solute carrier family 22 member 1 SLC22A1 HOCT1,
          OCT1,
          oct1_cds
          25.6kb Sense Chr. 6 - 160121789 - 160159201 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3410859 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P27415 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 31164 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28112 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3410859 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3410859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211263 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415201 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211260 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415154 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211257 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211255 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211246 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211199 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211200 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415193 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211204 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211269 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211210 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415163 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211259 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415193 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415163 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211199 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211257 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211260 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211200 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211210 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211259 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211246 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415154 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211263 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211255 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415201 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25415151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211204 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05211269 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P27415 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P27415 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211218 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211249 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211229 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211238 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211236 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211267 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211207 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211241 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211188 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211239 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211194 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211272 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211252 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211196 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15882866 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211187 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211221 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211266 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211247 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211223 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211182 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211215 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211179 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211271 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211261 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211220 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211205 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211259 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211243 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211185 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211204 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211227 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211226 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211274 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211269 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211192 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211186 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211254 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211234 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211273 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211231 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211264 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211199 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211210 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211202 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211256 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211212 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05211214 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_003288_at - -
          Affymetrix Human Gene 1.0 ST Array 8123231 - -
          Affymetrix Human Gene 1.1 ST Array 8123230 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032291 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032291 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 144924 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2180519 AK097332.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2180519 BC032536.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2180519 NR_003288.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2180519 AK096733.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2180519 AK097332.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2180519 BC032536.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2180519 NR_003288.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2180519 AK096733.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2180519 AK097332.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2180519 BC032536.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2180519 NR_003288.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2180519 AK096733.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2180519 AK097332.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2180519 BC032536.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2180519 NR_003288.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2180519 AK096733.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2180519 AK097332.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2180519 BC032536.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2180519 NR_003288.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2180519 AK096733.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2180519 AK097332.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2180519 BC032536.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2180519 NR_003288.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2180519 AK096733.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2180519 AK097332.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2180519 BC032536.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2180519 NR_003288.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2180519 AK096733.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2180519 AK097332.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2180519 BC032536.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2180519 NR_003288.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2180519 AK096733.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0