Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003309.3 | NP_003300.1 | 1 | 3730 | 84 | |
GenBank mRNA | AK096882.1 | - | 1 | 150 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TSPY like 4 | TSPYL4 | dJ486I3.2 | -20.8kb | Sense | Chr. 6 - 116249964 - 116254098 on NCBI Build GRCh38 |
Downstream | dermatan sulfate epimerase | DSE |
DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2 |
3.6kb | Anti-sense | Chr. 6 - 116254152 - 116441261 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |