Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657006_s1
          See other LOC646482 GE Assays ›
          Gene Symbol:
          LOC646482
          Gene Name:
          uncharacterized LOC646482
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657006_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC646482
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC015677.2 - 1 955 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream nucleoporin 58 NUP58 NUP45,
          NUPL1,
          PRO2463
          Anti-sense Chr. 13 - 25301152 - 25349795 on NCBI Build GRCh38
          Downstream ATPase phospholipid transporting 8A2 ATP8A2 ATP,
          ATPIB,
          CAMRQ4,
          IB,
          ML-1
          70.6kb Anti-sense Chr. 13 - 25372011 - 26025851 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 501842 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 328398 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3420879 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 17319 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3420879 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02739881 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1569517_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1300008389.hg.1 ENST00000568856.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300008389.hg.1 ENST00000568856.2 Ensembl
          Human Genome U133 Plus 2.0 Array 1569517_at - -
          Human Genome U133 Plus 2.0 Array 1569517_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569517_at5 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1799102 BC015677.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1653545 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1653545 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1799102 BC015677.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1799102 BC015677.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0