Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657160_s1
          See other LINC01003 GE Assays ›
          Gene Symbol:
          LINC01003
          Gene Name:
          long intergenic non-protein coding RNA 1003
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 152464124 - 152465545 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657160_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01003
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027387.1 1 1245 88
          GenBank mRNA BC036622.2 - 1 1150 88
          BU742627.1 - 1 88
          CF146980.1 - 1 591 88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI199521.1 - 1 Chr. 7 - 152465201 - 152465545 [-] on Build GRCh38 88
          GenBankmRNA DB574637.1 - 1 269 Chr. 7 - 152465100 - 152465543 [+] on Build GRCh38 88
          GenBankmRNA AI992275.1 - 1 Chr. 7 - 152465115 - 152465545 [-] on Build GRCh38 88
          GenBankmRNA BU079162.1 - 1 429 Chr. 7 - 152464943 - 152465427 [+] on Build GRCh38 88
          GenBankmRNA BU078878.1 - 1 Chr. 7 - 152465007 - 152465551 [-] on Build GRCh38 88
          GenBankmRNA AW207591.1 - 1 210 Chr. 7 - 152465159 - 152465545 [+] on Build GRCh38 88
          GenBankmRNA AA987524.1 - 1 192 Chr. 7 - 152465177 - 152465447 [+] on Build GRCh38 88
          GenBankmRNA BM728687.1 - 1 401 Chr. 7 - 152464968 - 152465545 [+] on Build GRCh38 88
          GenBankmRNA HY304546.1 - 1 Chr. 7 - 152465017 - 152465543 [-] on Build GRCh38 88
          GenBankmRNA AI220463.1 - 1 Chr. 7 - 152465152 - 152465544 [-] on Build GRCh38 88
          GenBankmRNA BU727559.1 - 1 Chr. 7 - 152465043 - 152465475 [-] on Build GRCh38 88
          GenBankmRNA AI458048.1 - 1 Chr. 7 - 152465040 - 152465475 [-] on Build GRCh38 88
          GenBankmRNA DB566301.1 - 1 291 Chr. 7 - 152465083 - 152465542 [+] on Build GRCh38 88
          GenBankmRNA AI277022.1 - 1 Chr. 7 - 152465303 - 152465544 [-] on Build GRCh38 88
          GenBankmRNA DB523302.1 - 1 306 Chr. 7 - 152465063 - 152465543 [+] on Build GRCh38 88
          GenBankmRNA HY352756.1 - Chr. - - [-] on Build 88
          GenBankmRNA HY302470.1 - 1 Chr. 7 - 152465057 - 152465543 [-] on Build GRCh38 88
          GenBankmRNA HY261138.1 - 1 Chr. 7 - 152465062 - 152465545 [-] on Build GRCh38 88
          GenBankmRNA AA404961.1 - 1 Chr. 7 - 152465206 - 152465545 [-] on Build GRCh38 88
          GenBankmRNA AA973506.1 - 1 192 Chr. 7 - 152465177 - 152465447 [+] on Build GRCh38 88
          GenBankmRNA AA431501.1 - 1 Chr. 7 - 152465262 - 152465545 [-] on Build GRCh38 88
          GenBankmRNA CK902549.1 - 1 336 Chr. 7 - 152465033 - 152465429 [+] on Build GRCh38 88
          GenBankmRNA HY270862.1 - 1 Chr. 7 - 152465101 - 152465543 [-] on Build GRCh38 88
          GenBankmRNA HY305110.1 - 1 Chr. 7 - 152465108 - 152465543 [-] on Build GRCh38 88
          GenBankmRNA BF195286.1 - 1 199 Chr. 7 - 152465170 - 152465446 [+] on Build GRCh38 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream fatty acid binding protein 5 pseudogene 3 FABP5P3 FABP5L3 -21.2kb Sense Chr. 7 - 152436895 - 152443016 on NCBI Build GRCh38
          Downstream X-ray repair cross complementing 2 XRCC2 FANCU 181.0kb Anti-sense Chr. 7 - 152646498 - 152676165 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700009740.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001028.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142297 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142282 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875849 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142299 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142311 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142280 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142278 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142306 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26142312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05875852 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 661648 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P6172 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P6172 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 19738 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18227 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P6172 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P6172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875833 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875862 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875849 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875847 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142299 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875844 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875843 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875839 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875826 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142298 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875837 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875851 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875827 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142273 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875835 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142262 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875835 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875839 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142273 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875826 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875833 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875827 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875843 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875849 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875862 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142299 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142298 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875851 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875844 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875837 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142262 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875847 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P6172 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16331493 - -
          Agilent Human 1B Microarray G4111A A_32_P6172 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875825 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875850 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875852 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875847 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875848 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875846 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875834 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875877 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875868 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16040320 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875861 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875876 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875835 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16040319 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875871 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875864 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875874 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875859 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875839 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875857 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16040317 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875855 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16040322 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875832 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875851 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875843 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16040314 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875838 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875866 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875854 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875829 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05875872 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 235174_PM_s_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR07015925.hg.1 NR_027387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07015925.hg.1 TC07001028.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700009740.hg.1 ENST00000564834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009740.hg.1 ENST00000564834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009740.hg.1 NR_027387.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009740.hg.1 ENST00000564834.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009740.hg.1 ENST00000564834.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009740.hg.1 NR_027387.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169516.hg.1 ENST00000564834.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169516.hg.1 NR_027387.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169516.hg.1 TC0700009740.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700169516.hg.1 NR_027387.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169516.hg.1 ENST00000564834.1 Ensembl
          Human Genome U133 Plus 2.0 Array 235174_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 235174_s_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235174_s_at6 - -
          Human Genome U95 Set 44567_at - -
          Human Genome U95 Set 44567_PM_at - -
          Human Genome U133 Set 235174_PM_s_at - -
          Human Genome U133 Set 235174_s_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035366 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035366 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3195658 - -
          HumanHT-12 v4 Expression BeadChip ILMN_3272129 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0