Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657277_m1
          See other LOC284570 GE Assays ›
          Gene Symbol:
          LOC284570
          Gene Name:
          uncharacterized LOC284570
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657277_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC284570
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC040156.1 - 1 53 94
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG910726.1 - 1 53 Chr. 1 - 202130056 - 202130905 [+] on Build GRCh38 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream G protein-coupled receptor 37 like 1 GPR37L1 ET(B)R-LP-2,
          ETBR-LP-2
          Sense Chr. 1 - 202118669 - 202133588 on NCBI Build GRCh38
          Downstream ADP ribosylation factor like GTPase 8A ARL8A ARL10B,
          GIE2
          2.0kb Anti-sense Chr. 1 - 202133404 - 202144743 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Amplicon spans exons and probe does not span exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3469658 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P114866 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 23093 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43235 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3469658 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P114866 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P114866 - -
          Agilent Human 1B Microarray G4111A A_32_P114866 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1558189_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 NM_004767.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 NM_004767.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 NM_004767.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011211.hg.1 NM_004767.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 ENST00000367282.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 NM_004767.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 NM_004767.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 NM_004767.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011211.hg.1 NM_004767.4 Refseq
          Human Genome U133 Plus 2.0 Array 1558189_a_at - -
          Human Genome U133 Plus 2.0 Array 1558189_PM_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558189_a_at1 - -
          Human Genome U95 Set 88190_PM_at - -
          Human Genome U95 Set 88190_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0