Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657382_m1
          See other LOC440028 GE Assays ›
          Gene Symbol:
          LOC440028
          Gene Name:
          uncharacterized LOC440028
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 9754770 - 9759533 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657382_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC440028
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033972.1 2 - 3 185 105
          GenBank mRNA AK090613.1 - 2 - 3 185 105
          BC040853.1 - 1 - 2 401 105
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM479379.1 - 1 - 2 401 Chr. 11 - 9756378 - 9758164 [-] on Build GRCh38 105
          GenBankmRNA DA047764.1 - 2 - 3 185 Chr. 11 - 9756547 - 9759533 [-] on Build GRCh38 105

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SWAP switching B-cell complex 70kDa subunit SWAP70 HSPC321,
          SWAP-70
          -2.1kb Anti-sense Chr. 11 - 9664077 - 9752991 on NCBI Build GRCh38
          Downstream SBF2 antisense RNA 1 SBF2-AS1 129.0 Anti-sense Chr. 11 - 9758293 - 9811319 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100010075.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001406.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 163229 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 446769 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 96952 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3833256 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 51573 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02390832 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3833256 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3833256 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02390832 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02390828 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02390832 - -
          Agilent Human 1B Microarray G4111A A_32_P904828 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02390832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1562606_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11017692.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11017692.hg.1 NR_033972.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11017692.hg.1 TC11001406.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11017694.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11017694.hg.1 NR_033972.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11017694.hg.1 TC11001406.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11017693.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11017693.hg.1 NR_033972.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11017693.hg.1 TC11001406.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100176357.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176357.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176357.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176357.hg.1 TC1100010075.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100176357.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176357.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176357.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176356.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176356.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176356.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176356.hg.1 TC1100010075.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100176356.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176356.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176356.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176352.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176352.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176352.hg.1 TC1100010075.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100176352.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176352.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176354.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176354.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176354.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176354.hg.1 TC1100010075.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100176354.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176354.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176354.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176353.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176353.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176353.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176353.hg.1 TC1100010075.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100176353.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176353.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176353.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176360.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176360.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100176360.hg.1 TC1100010075.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100176360.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100176360.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000500698.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 ENST00000525154.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 NR_033972.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010075.hg.1 NR_033972.1 Refseq
          Human Genome U133 Plus 2.0 Array 1566830_at - -
          Human Genome U133 Plus 2.0 Array 1562606_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 1562606_a_at - -
          Human Genome U133 Plus 2.0 Array 1566830_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562606_a_at2 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044930 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044930 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0