Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657701_m1
          See other LOC338694 GE Assays ›
          Gene Symbol:
          LOC338694
          Gene Name:
          uncharacterized LOC338694
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 69147228 - 69171562 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657701_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC338694
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104161.1 2 - 3 480 83
          GenBank mRNA BC043531.1 - 2 - 3 480 83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI826779.1 - 2 - 3 482 Chr. 11 - 69147386 - 69171564 [-] on Build GRCh38 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 3164 MIR3164 -64.0kb Anti-sense Chr. 11 - 69083176 - 69083258 on NCBI Build GRCh38
          Downstream uncharacterized LOC105369367 LOC105369367 60.3kb Sense Chr. 11 - 69228749 - 69234936 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100011421.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856613 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28272183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856648 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856636 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28272026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856635 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28272157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28272176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856624 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856630 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28272141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856633 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856620 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28272146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16467728 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856642 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856615 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28272171 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28272140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28272019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856637 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16467727 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28272147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07856578 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 324061 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 570996 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3556839 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60025 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3556839 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02463275 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02463269 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1563042_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100011421.hg.1 ENST00000562772.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011421.hg.1 ENST00000562772.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011421.hg.1 ENST00000562772.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011421.hg.1 NR_104161.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100011421.hg.1 NR_104161.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100011421.hg.1 NR_104161.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100188082.hg.1 ENST00000562772.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188082.hg.1 NR_104161.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100188082.hg.1 TC1100011421.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100188082.hg.1 ENST00000562772.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188082.hg.1 NR_104161.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011421.hg.1 ENST00000562772.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011421.hg.1 ENST00000562772.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011421.hg.1 ENST00000562772.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011421.hg.1 NR_104161.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011421.hg.1 NR_104161.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011421.hg.1 NR_104161.1 Refseq
          Human Genome U133 Plus 2.0 Array 1563042_at - -
          Human Genome U133 Plus 2.0 Array 1563042_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563042_at6 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1853834 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 6040288 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_116933 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1853834 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1853834 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1853834 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0