Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03658752_s1
          See other LOC642648 GE Assays ›
          Gene Symbol:
          LOC642648
          Gene Name:
          translation initiation factor IF-2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.22: 46067464 - 46071470 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03658752_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC642648
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK123383.1 - 1 2052 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB306980.1 - 1 Chr. 22 - 46068721 - 46069262 [+] on Build GRCh38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream PRR34 antisense RNA 1 PRR34-AS1 -10.2kb Anti-sense Chr. 22 - 46053846 - 46058522 on NCBI Build GRCh38
          Downstream MIRLET7B host gene MIRLET7BHG linc-Ppara 15.0kb Anti-sense Chr. 22 - 46085997 - 46113928 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219105 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219119 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219090 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219109 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219094 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219082 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219112 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219088 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219127 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219093 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11530139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32219091 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3267431 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55932 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3267431 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_041270_at - -
          Affymetrix Human Gene 1.0 ST Array 8076748 - -
          Affymetrix Human Gene 1.1 ST Array 8076747 - -
          Affymetrix Human Transcriptome Array 2.0 TC2200009002.hg.1 ENST00000412643.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009002.hg.1 ENST00000412643.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009002.hg.1 ENST00000412643.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009002.hg.1 ENST00000412643.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009002.hg.1 ENST00000412643.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009002.hg.1 ENST00000412643.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1682335 AK123383.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1682335 AK123383.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1682335 AK123383.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1682335 AK123383.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1682335 AK123383.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0