Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03659553_s1
          See other LOC100127909 GE Assays ›
          Gene Symbol:
          LOC100127909
          Gene Name:
          uncharacterized LOC100127909
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03659553_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100127909
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK130422.1 - 1 555 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger and BTB domain containing 7C ZBTB7C APM-1,
          APM1,
          ZBTB36,
          ZNF857C
          Sense Chr. 18 - 48026886 - 48412639 on NCBI Build GRCh38
          Downstream chromosome 18 open reading frame 12 C18orf12 HEIL1,
          HsT2508
          117.2kb Anti-sense Chr. 18 - 48252301 - 48253185 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31291397 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17066088 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31291384 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31291394 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17066085 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17066090 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17066087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31291382 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31291390 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10690587 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 25155 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3398181 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3179038 AK130422.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0