Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03659807_m1
          See other LOC389273 GE Assays ›
          Gene Symbol:
          LOC389273
          Gene Name:
          uncharacterized LOC389273
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 10504990 - 10522084 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03659807_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC389273
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_134289.1 2 - 3 433 80
          NR_134290.1 2 - 3 242 80
          GenBank mRNA AK130861.1 - 2 - 3 242 80
          CR740839.1 - 2 - 3 433 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC101929412 LOC101929412 -2.3kb Sense Chr. 5 - 10493527 - 10502728 on NCBI Build GRCh38
          Downstream ankyrin repeat domain 33B ANKRD33B 57.7kb Anti-sense Chr. 5 - 10564323 - 10657816 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23927673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23927650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23927636 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762171 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762167 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23927671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23927654 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23927656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762137 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23927648 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03762170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23927643 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 221381 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3430391 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 3683 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3430391 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37087140 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0500010093.hg.1 ENST00000504650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010093.hg.1 ENST00000504650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010093.hg.1 ENST00000504650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010093.hg.1 ENST00000506021.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010093.hg.1 ENST00000506021.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010093.hg.1 ENST00000506021.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010093.hg.1 ENST00000506021.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010093.hg.1 ENST00000504650.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010093.hg.1 ENST00000504650.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010093.hg.1 ENST00000504650.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010093.hg.1 ENST00000506021.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010093.hg.1 ENST00000506021.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010093.hg.1 ENST00000506021.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010093.hg.1 ENST00000506021.4 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1855587 AK130861.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1855587 AK130861.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1855587 AK130861.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_99570 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_99570 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1855587 AK130861.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1855587 AK130861.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1855587 AK130861.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0