Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03660343_s1
          See other CNKSR2 GE Assays ›
          Gene Symbol:
          CNKSR2
          Gene Name:
          connector enhancer of kinase suppressor of Ras 2
          Gene Aliases:
          CNK2, KSR2, MAGUIN
          Chromosome Location:
          Chr.X: 21374335 - 21654695 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03660343_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CNKSR2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001168647.1 NP_001162118.1 21 5031 66
          NM_014927.3 NP_055742.2 22 5121 66
          GenBank mRNA AK124258.1 - 1 1655 66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW468132.1 - 1 Chr. X - 21653860 - 21654272 [-] on Build GRCh38 66
          GenBankmRNA BF432843.1 - 1 Chr. X - 21653771 - 21654280 [-] on Build GRCh38 66
          GenBankmRNA H52132.1 - Chr. - - [-] on Build 66
          GenBankmRNA AA514014.1 - 1 Chr. X - 21653941 - 21654286 [-] on Build GRCh38 66
          GenBankmRNA AI670947.1 - 1 Chr. X - 21653652 - 21654272 [-] on Build GRCh38 66
          GenBankmRNA AW972829.1 - 1 Chr. X - 21653659 - 21654293 [-] on Build GRCh38 66
          GenBankmRNA AI361550.1 - 1 Chr. X - 21653874 - 21654273 [-] on Build GRCh38 66
          GenBankmRNA AW003080.1 - 1 Chr. X - 21653800 - 21654272 [-] on Build GRCh38 66
          GenBankmRNA BE674495.1 - 1 Chr. X - 21653756 - 21654275 [-] on Build GRCh38 66
          GenBankmRNA AW900550.1 - 1 Chr. X - 21653835 - 21654240 [-] on Build GRCh38 66
          GenBankmRNA AW237040.1 - 1 Chr. X - 21653710 - 21654272 [-] on Build GRCh38 66
          GenBankmRNA BE670298.1 - 1 Chr. X - 21653825 - 21654275 [-] on Build GRCh38 66

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ribosomal protein S6 kinase A3 RPS6KA3 CLS,
          HU-3,
          ISPK-1,
          MAPKAPK1B,
          MRX19,
          RSK,
          RSK2,
          S6K-alpha3,
          p90-RSK2,
          pp90RSK2
          -1.4mb Anti-sense Chr. X - 20149911 - 20267514 on NCBI Build GRCh38
          Downstream kelch like family member 34 KLHL34 662.0 Anti-sense Chr. X - 21655349 - 21658387 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CNKSR2

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of signal transduction
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0