Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03660905_s1
          See other LOC100131654 GE Assays ›
          Gene Symbol:
          LOC100131654
          Gene Name:
          uncharacterized LOC100131654
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03660905_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100131654
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK124891.1 - 1 951 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB309585.1 - 1 Chr. 15 - 89393448 - 89393989 [-] on Build GRCh38 80
          GenBankmRNA AW293809.1 - 1 Chr. 15 - 89393443 - 89393850 [-] on Build GRCh38 80
          GenBankmRNA AI494385.1 - 1 Chr. 15 - 89393433 - 89393836 [-] on Build GRCh38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 9-3 MIR9-3 MIRN9-3,
          hsa-mir-9-3,
          miRNA9-3,
          mir-9-3
          -24.6kb Sense Chr. 15 - 89368017 - 89368106 on NCBI Build GRCh38
          Downstream translation initiation factor IF-2-like LOC105371031 6.1kb Sense Chr. 15 - 89399716 - 89507395 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16825275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820432 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820440 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820439 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16825270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324459 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16825272 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16825273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820420 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16825277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324447 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820433 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820437 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820429 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820411 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324407 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820425 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820423 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16825279 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820451 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820406 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820426 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820407 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820405 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820421 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324416 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16825282 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820448 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324422 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30324405 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09820446 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3253465 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 643 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3253465 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0