Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000657.hg.1 |
ENST00000399868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000657.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000657.hg.1 |
TC10000057.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000658.hg.1 |
ENST00000399868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000658.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000658.hg.1 |
TC10000057.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000656.hg.1 |
ENST00000399868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000656.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000656.hg.1 |
TC10000057.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000145680.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145680.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145680.hg.1 |
TC1000006664.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000145680.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145680.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145679.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145679.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145679.hg.1 |
TC1000006664.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000145679.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145679.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145681.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145681.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145681.hg.1 |
TC1000006664.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000145681.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145681.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
ENST00000399868.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006664.hg.1 |
NR_040079.2 |
Refseq |