Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03664089_s1
          See other LOC100129186 GE Assays ›
          Gene Symbol:
          LOC100129186
          Gene Name:
          uncharacterized LOC100129186
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03664089_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100129186
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK128204.1 - 1 2947 65
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF868540.1 - 1 319 Chr. 5 - 42954063 - 42954477 [+] on Build GRCh38 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream selenoprotein P,
          plasma,
          1
          SEPP1 SELP,
          SEPP,
          SeP
          -139.5kb Anti-sense Chr. 5 - 42799880 - 42811922 on NCBI Build GRCh38
          Downstream uncharacterized LOC643977 FLJ32255 30.4kb Anti-sense Chr. 5 - 42985398 - 42993333 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902166 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074127 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074201 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15686674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902202 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074144 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15686685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24074128 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03902190 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 242739 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3342967 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3363460 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 37965 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 46643 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3342967 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3363460 - -
          Agilent Human 1B Microarray G4111A A_32_P536941 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3265193 AK128204.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3187210 AK128204.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0